Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 56389247-56389508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX9
Entrez Gene ID
374618
Gene Name
testis expressed 9
Gene Aliases
-
UniGene
Hs.511476
Gene Chromosome Location
Chr.15: 56243971-56445997 on Build GRCh38
Transcripts
NM_001286449.1 NM_198524.2 XM_005254359.4 XM_005254361.3 XM_011521530.2 XM_017022160.1 XM_017022161.1 XM_017022162.1 XM_017022163.1 XM_017022164.1 XM_017022166.1 XM_017022167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427986 c.315A>G p.P105P Chr.15: 56389320-56389320 on Build GRCh38
5369341 c.338T>C p.V113A Chr.15: 56389343-56389343 on Build GRCh38
6077634 c.395+1G>A p.? Chr.15: 56389401-56389401 on Build GRCh38
963213 c.390T>C p.N130N Chr.15: 56389395-56389395 on Build GRCh38
1723314 c.370A>G p.N124D Chr.15: 56389375-56389375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759389003 Chr.15: 56389353-56389353 on Build GRCh38
rs564181683 Chr.15: 56389370-56389370 on Build GRCh38
rs758894571 Chr.15: 56389401-56389401 on Build GRCh38
rs750952594 Chr.15: 56389378-56389378 on Build GRCh38
rs769310757 Chr.15: 56389425-56389425 on Build GRCh38
rs931342777 Chr.15: 56389344-56389344 on Build GRCh38
rs751970915 Chr.15: 56389403-56389403 on Build GRCh38
rs773061492 Chr.15: 56389426-56389427 on Build GRCh38
rs777076681 Chr.15: 56389435-56389435 on Build GRCh38
rs780873252 Chr.15: 56389407-56389408 on Build GRCh38
rs779513962 Chr.15: 56389287-56389287 on Build GRCh38
rs780598936 Chr.15: 56389402-56389402 on Build GRCh38
rs141431012 Chr.15: 56389427-56389427 on Build GRCh38
rs947978679 Chr.15: 56389285-56389285 on Build GRCh38
rs544519606 Chr.15: 56389313-56389313 on Build GRCh38
rs755307419 Chr.15: 56389408-56389408 on Build GRCh38
rs746592659 Chr.15: 56389350-56389350 on Build GRCh38
rs917734935 Chr.15: 56389281-56389281 on Build GRCh38
rs1008296191 Chr.15: 56389475-56389475 on Build GRCh38
rs898501931 Chr.15: 56389336-56389336 on Build GRCh38
rs374231217 Chr.15: 56389294-56389294 on Build GRCh38
rs747860136 Chr.15: 56389426-56389426 on Build GRCh38
rs764458362 Chr.15: 56389354-56389354 on Build GRCh38
rs747753055 Chr.15: 56389422-56389422 on Build GRCh38
rs186349415 Chr.15: 56389334-56389334 on Build GRCh38
rs774077123 Chr.15: 56389340-56389340 on Build GRCh38
rs780216497 Chr.15: 56389295-56389295 on Build GRCh38
rs746320944 Chr.15: 56389293-56389293 on Build GRCh38
rs200296454 Chr.15: 56389359-56389359 on Build GRCh38
rs28369022 Chr.15: 56389300-56389300 on Build GRCh38
rs765181178 Chr.15: 56389375-56389375 on Build GRCh38
rs768267414 Chr.15: 56389378-56389381 on Build GRCh38
rs757443034 Chr.15: 56389372-56389372 on Build GRCh38
rs747699751 Chr.15: 56389302-56389302 on Build GRCh38
rs377237972 Chr.15: 56389296-56389296 on Build GRCh38
rs762745230 Chr.15: 56389327-56389327 on Build GRCh38
rs780857020 Chr.15: 56389409-56389409 on Build GRCh38

View Full Product Details