Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21536312-21536553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF429
Entrez Gene ID
353088
Gene Name
zinc finger protein 429
Gene Aliases
-
UniGene
Hs.572567
Gene Chromosome Location
Chr.19: 21505257-21541157 on Build GRCh38
Transcripts
NM_001001415.2 XM_017026748.1 XM_017026749.1 XM_017026750.1 XM_017026751.1 XM_017026752.1 XM_017026753.1 XM_017026754.1 XM_017026755.1 XM_017026756.1 XM_017026757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982237 c.437A>G p.H146R Chr.19: 21536490-21536490 on Build GRCh38
4973510 c.439T>C p.C147R Chr.19: 21536492-21536492 on Build GRCh38
4076190 c.426C>T p.S142S Chr.19: 21536479-21536479 on Build GRCh38
3748673 c.415C>A p.L139I Chr.19: 21536468-21536468 on Build GRCh38
993692 c.447A>G p.I149M Chr.19: 21536500-21536500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774404331 Chr.19: 21536423-21536423 on Build GRCh38
rs1034986417 Chr.19: 21536515-21536515 on Build GRCh38
rs777474414 Chr.19: 21536485-21536485 on Build GRCh38
rs943624910 Chr.19: 21536410-21536410 on Build GRCh38
rs747799527 Chr.19: 21536467-21536470 on Build GRCh38
rs199734275 Chr.19: 21536488-21536488 on Build GRCh38
rs1011847537 Chr.19: 21536430-21536430 on Build GRCh38
rs748813439 Chr.19: 21536360-21536360 on Build GRCh38
rs751029456 Chr.19: 21536401-21536408 on Build GRCh38
rs776519889 Chr.19: 21536521-21536521 on Build GRCh38
rs201746630 Chr.19: 21536433-21536433 on Build GRCh38
rs557054437 Chr.19: 21536449-21536449 on Build GRCh38
rs570892795 Chr.19: 21536439-21536439 on Build GRCh38
rs537821409 Chr.19: 21536520-21536520 on Build GRCh38
rs773008812 Chr.19: 21536378-21536378 on Build GRCh38
rs751864922 Chr.19: 21536346-21536346 on Build GRCh38
rs762927518 Chr.19: 21536415-21536415 on Build GRCh38
rs773854622 Chr.19: 21536437-21536437 on Build GRCh38
rs764232949 Chr.19: 21536339-21536339 on Build GRCh38
rs796937259 Chr.19: 21536373-21536373 on Build GRCh38
rs762143911 Chr.19: 21536428-21536428 on Build GRCh38
rs757556696 Chr.19: 21536348-21536348 on Build GRCh38
rs372280271 Chr.19: 21536479-21536479 on Build GRCh38
rs370582019 Chr.19: 21536370-21536370 on Build GRCh38
rs62108336 Chr.19: 21536411-21536411 on Build GRCh38
rs148845371 Chr.19: 21536442-21536442 on Build GRCh38
rs375806784 Chr.19: 21536517-21536517 on Build GRCh38
rs111767938 Chr.19: 21536362-21536362 on Build GRCh38
rs750850967 Chr.19: 21536436-21536436 on Build GRCh38
rs138757574 Chr.19: 21536380-21536380 on Build GRCh38
rs770856473 Chr.19: 21536403-21536403 on Build GRCh38
rs756493581 Chr.19: 21536417-21536417 on Build GRCh38
rs764299781 Chr.19: 21536421-21536421 on Build GRCh38
rs781650441 Chr.19: 21536352-21536352 on Build GRCh38
rs778420112 Chr.19: 21536363-21536363 on Build GRCh38
rs758974432 Chr.19: 21536450-21536450 on Build GRCh38
rs758012235 Chr.19: 21536485-21536486 on Build GRCh38
rs747905615 Chr.19: 21536369-21536369 on Build GRCh38
rs114046517 Chr.19: 21536472-21536472 on Build GRCh38
rs780573587 Chr.19: 21536426-21536426 on Build GRCh38
rs375919575 Chr.19: 21536359-21536359 on Build GRCh38
rs373172333 Chr.19: 21536355-21536355 on Build GRCh38
rs200958386 Chr.19: 21536460-21536460 on Build GRCh38

View Full Product Details