Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43847351-43847580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF283
Entrez Gene ID
284349
Gene Name
zinc finger protein 283
Gene Aliases
HZF19, HZF41
UniGene
Hs.652513
Gene Chromosome Location
Chr.19: 43827292-43853286 on Build GRCh38
Transcripts
NM_001297752.1 NM_181845.1 XM_005258784.3 XM_005258785.3 XM_005258786.3 XM_017026628.1 XM_017026629.1 XM_017026630.1 XM_017026631.1 XM_017026632.1 XM_017026633.1 XM_017026634.1 XM_017026635.1 XM_017026636.1 XM_017026637.1 XM_017026638.1 XM_017026639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
997721 c.848G>T p.R283I Chr.19: 43847449-43847449 on Build GRCh38
6290640 c.861T>A p.G287G Chr.19: 43847462-43847462 on Build GRCh38
997720 c.776G>A p.G259D Chr.19: 43847377-43847377 on Build GRCh38
4132005 c.941C>T p.T314I Chr.19: 43847542-43847542 on Build GRCh38
4900064 c.853C>T p.H285Y Chr.19: 43847454-43847454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746998210 Chr.19: 43847540-43847540 on Build GRCh38
rs373649379 Chr.19: 43847527-43847527 on Build GRCh38
rs776131017 Chr.19: 43847472-43847472 on Build GRCh38
rs745663648 Chr.19: 43847442-43847442 on Build GRCh38
rs752724552 Chr.19: 43847510-43847510 on Build GRCh38
rs865863615 Chr.19: 43847454-43847454 on Build GRCh38
rs750754292 Chr.19: 43847483-43847483 on Build GRCh38
rs780115858 Chr.19: 43847537-43847537 on Build GRCh38
rs774129864 Chr.19: 43847433-43847433 on Build GRCh38
rs758797866 Chr.19: 43847492-43847492 on Build GRCh38
rs764798445 Chr.19: 43847477-43847477 on Build GRCh38
rs368697755 Chr.19: 43847506-43847506 on Build GRCh38
rs752112464 Chr.19: 43847503-43847503 on Build GRCh38
rs763744692 Chr.19: 43847469-43847469 on Build GRCh38
rs762934749 Chr.19: 43847451-43847451 on Build GRCh38
rs772031093 Chr.19: 43847528-43847528 on Build GRCh38
rs761590446 Chr.19: 43847476-43847476 on Build GRCh38
rs372529241 Chr.19: 43847378-43847378 on Build GRCh38
rs570503505 Chr.19: 43847377-43847377 on Build GRCh38
rs2195980 Chr.19: 43847542-43847542 on Build GRCh38
rs775599790 Chr.19: 43847452-43847452 on Build GRCh38
rs766842656 Chr.19: 43847498-43847498 on Build GRCh38
rs748462375 Chr.19: 43847389-43847389 on Build GRCh38
rs777879571 Chr.19: 43847512-43847512 on Build GRCh38
rs376591773 Chr.19: 43847435-43847435 on Build GRCh38
rs755039644 Chr.19: 43847505-43847505 on Build GRCh38
rs878967874 Chr.19: 43847548-43847548 on Build GRCh38
rs982404307 Chr.19: 43847447-43847447 on Build GRCh38
rs781373508 Chr.19: 43847386-43847386 on Build GRCh38
rs952440443 Chr.19: 43847423-43847423 on Build GRCh38
rs371059248 Chr.19: 43847523-43847523 on Build GRCh38
rs879241206 Chr.19: 43847525-43847525 on Build GRCh38
rs760229541 Chr.19: 43847456-43847456 on Build GRCh38
rs372629226 Chr.19: 43847414-43847414 on Build GRCh38

View Full Product Details