Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19114944-19115211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPRIPL2
Entrez Gene ID
162073
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Gene Aliases
-
UniGene
Hs.530899
Gene Chromosome Location
Chr.16: 19113932-19121630 on Build GRCh38
Transcripts
NM_001034841.3 NR_028028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7219541 c.490G>A p.E164K Chr.16: 19114951-19114951 on Build GRCh38
5181325 c.663delG p.G223fs*56 Chr.16: 19115124-19115124 on Build GRCh38
3506656 c.650C>T p.P217L Chr.16: 19115111-19115111 on Build GRCh38
3402121 c.550C>G p.P184A Chr.16: 19115011-19115011 on Build GRCh38
4693637 c.712G>A p.V238M Chr.16: 19115173-19115173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776927294 Chr.16: 19115152-19115152 on Build GRCh38
rs202102935 Chr.16: 19115186-19115186 on Build GRCh38
rs763147007 Chr.16: 19115119-19115119 on Build GRCh38
rs377492063 Chr.16: 19115173-19115173 on Build GRCh38
rs762495729 Chr.16: 19115172-19115172 on Build GRCh38
rs763928448 Chr.16: 19115067-19115067 on Build GRCh38
rs769870640 Chr.16: 19115048-19115048 on Build GRCh38
rs372459531 Chr.16: 19114974-19114974 on Build GRCh38
rs763070979 Chr.16: 19115056-19115056 on Build GRCh38
rs143405390 Chr.16: 19115094-19115094 on Build GRCh38
rs769030900 Chr.16: 19115020-19115020 on Build GRCh38
rs753166483 Chr.16: 19115189-19115189 on Build GRCh38
rs1003851344 Chr.16: 19115128-19115128 on Build GRCh38
rs778516824 Chr.16: 19114991-19114991 on Build GRCh38
rs767856490 Chr.16: 19115188-19115188 on Build GRCh38
rs776497709 Chr.16: 19115144-19115144 on Build GRCh38
rs116496443 Chr.16: 19115148-19115148 on Build GRCh38
rs149205864 Chr.16: 19115093-19115094 on Build GRCh38
rs8051801 Chr.16: 19115170-19115170 on Build GRCh38
rs756810996 Chr.16: 19114978-19114978 on Build GRCh38
rs567647380 Chr.16: 19115125-19115125 on Build GRCh38
rs746989161 Chr.16: 19115120-19115120 on Build GRCh38
rs761530962 Chr.16: 19115071-19115071 on Build GRCh38
rs547667965 Chr.16: 19115077-19115077 on Build GRCh38
rs755387658 Chr.16: 19115085-19115085 on Build GRCh38
rs550861639 Chr.16: 19115081-19115081 on Build GRCh38
rs200640643 Chr.16: 19115009-19115009 on Build GRCh38
rs145081579 Chr.16: 19115100-19115100 on Build GRCh38
rs999860568 Chr.16: 19114970-19114970 on Build GRCh38
rs760428343 Chr.16: 19115066-19115066 on Build GRCh38
rs771105140 Chr.16: 19115111-19115111 on Build GRCh38
rs200493379 Chr.16: 19115088-19115088 on Build GRCh38
rs752080967 Chr.16: 19115083-19115083 on Build GRCh38
rs115342574 Chr.16: 19115181-19115181 on Build GRCh38
rs772196537 Chr.16: 19115058-19115058 on Build GRCh38
rs865805011 Chr.16: 19115092-19115093 on Build GRCh38
rs749867202 Chr.16: 19115003-19115003 on Build GRCh38
rs770072202 Chr.16: 19115089-19115089 on Build GRCh38
rs780523773 Chr.16: 19115005-19115005 on Build GRCh38
rs775565264 Chr.16: 19115063-19115063 on Build GRCh38
rs768410244 Chr.16: 19115134-19115134 on Build GRCh38
rs748429411 Chr.16: 19115037-19115037 on Build GRCh38
rs756544816 Chr.16: 19115191-19115191 on Build GRCh38
rs781314842 Chr.16: 19115022-19115022 on Build GRCh38
rs773432951 Chr.16: 19115054-19115054 on Build GRCh38
rs764811485 Chr.16: 19115076-19115076 on Build GRCh38
rs764453154 Chr.16: 19115069-19115070 on Build GRCh38
rs749447918 Chr.16: 19115101-19115101 on Build GRCh38
rs753536088 Chr.16: 19115070-19115070 on Build GRCh38
rs1048111990 Chr.16: 19115012-19115012 on Build GRCh38
rs912451358 Chr.16: 19114967-19114967 on Build GRCh38

View Full Product Details