Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41900407-41900680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3 XM_017022100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
961630 c.1596G>A p.S532S Chr.15: 41900675-41900675 on Build GRCh38
6652446 c.1602G>T p.R534S Chr.15: 41900669-41900669 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897037053 Chr.15: 41900592-41900592 on Build GRCh38
rs747406321 Chr.15: 41900642-41900642 on Build GRCh38
rs953873763 Chr.15: 41900616-41900616 on Build GRCh38
rs777215958 Chr.15: 41900610-41900610 on Build GRCh38
rs555780580 Chr.15: 41900457-41900457 on Build GRCh38
rs1002683793 Chr.15: 41900566-41900566 on Build GRCh38
rs1030527134 Chr.15: 41900445-41900445 on Build GRCh38
rs778505586 Chr.15: 41900627-41900627 on Build GRCh38
rs775988621 Chr.15: 41900634-41900634 on Build GRCh38
rs950342854 Chr.15: 41900528-41900528 on Build GRCh38
rs919046482 Chr.15: 41900525-41900525 on Build GRCh38
rs763343727 Chr.15: 41900661-41900661 on Build GRCh38
rs937902352 Chr.15: 41900559-41900559 on Build GRCh38
rs560617600 Chr.15: 41900522-41900522 on Build GRCh38
rs748154378 Chr.15: 41900436-41900436 on Build GRCh38
rs946611219 Chr.15: 41900589-41900589 on Build GRCh38
rs924168006 Chr.15: 41900432-41900432 on Build GRCh38
rs370652136 Chr.15: 41900650-41900650 on Build GRCh38
rs200600029 Chr.15: 41900628-41900628 on Build GRCh38
rs903189587 Chr.15: 41900432-41900432 on Build GRCh38
rs552787030 Chr.15: 41900555-41900555 on Build GRCh38
rs761925809 Chr.15: 41900663-41900663 on Build GRCh38
rs981800083 Chr.15: 41900546-41900546 on Build GRCh38
rs936931143 Chr.15: 41900435-41900435 on Build GRCh38
rs756909109 Chr.15: 41900624-41900624 on Build GRCh38
rs112415087 Chr.15: 41900586-41900586 on Build GRCh38
rs954931063 Chr.15: 41900455-41900455 on Build GRCh38
rs574286230 Chr.15: 41900551-41900551 on Build GRCh38
rs544194547 Chr.15: 41900593-41900593 on Build GRCh38
rs1043105085 Chr.15: 41900591-41900591 on Build GRCh38
rs915024473 Chr.15: 41900575-41900575 on Build GRCh38
rs531962895 Chr.15: 41900611-41900611 on Build GRCh38
rs773716100 Chr.15: 41900484-41900484 on Build GRCh38
rs367676526 Chr.15: 41900574-41900574 on Build GRCh38
rs963588731 Chr.15: 41900511-41900511 on Build GRCh38
rs1007831074 Chr.15: 41900463-41900463 on Build GRCh38
rs139614532 Chr.15: 41900651-41900651 on Build GRCh38
rs948152371 Chr.15: 41900542-41900542 on Build GRCh38

View Full Product Details