Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74798877-74799136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSK
Entrez Gene ID
1445
Gene Name
c-src tyrosine kinase
Gene Aliases
-
UniGene
Hs.77793
Gene Chromosome Location
Chr.15: 74782064-74803198 on Build GRCh38
Transcripts
NM_001127190.1 NM_004383.2 XM_005254165.4 XM_017021925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1938800 c.181C>T p.P61S Chr.15: 74798877-74798877 on Build GRCh38
3503992 c.182C>T p.P61L Chr.15: 74798878-74798878 on Build GRCh38
6211111 c.242+7A>C p.? Chr.15: 74798945-74798945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749484232 Chr.15: 74798969-74798969 on Build GRCh38
rs777445984 Chr.15: 74798951-74798951 on Build GRCh38
rs936483388 Chr.15: 74799075-74799075 on Build GRCh38
rs768133269 Chr.15: 74798918-74798918 on Build GRCh38
rs751293571 Chr.15: 74798955-74798955 on Build GRCh38
rs368457981 Chr.15: 74798930-74798930 on Build GRCh38
rs933223458 Chr.15: 74799101-74799101 on Build GRCh38
rs866432283 Chr.15: 74799011-74799011 on Build GRCh38
rs994838656 Chr.15: 74798927-74798927 on Build GRCh38
rs567254276 Chr.15: 74799064-74799064 on Build GRCh38
rs764983353 Chr.15: 74798933-74798933 on Build GRCh38
rs11553760 Chr.15: 74798906-74798906 on Build GRCh38
rs74607979 Chr.15: 74798957-74798957 on Build GRCh38
rs752325756 Chr.15: 74798949-74798949 on Build GRCh38
rs1025056010 Chr.15: 74799061-74799061 on Build GRCh38
rs759095664 Chr.15: 74798931-74798931 on Build GRCh38
rs768855120 Chr.15: 74798970-74798970 on Build GRCh38
rs775729597 Chr.15: 74798946-74798947 on Build GRCh38
rs1028108201 Chr.15: 74798946-74798946 on Build GRCh38
rs781081953 Chr.15: 74798958-74798958 on Build GRCh38
rs372362241 Chr.15: 74798950-74798950 on Build GRCh38
rs960572145 Chr.15: 74799033-74799033 on Build GRCh38
rs570998192 Chr.15: 74799032-74799033 on Build GRCh38
rs780130761 Chr.15: 74798967-74798967 on Build GRCh38
rs774922858 Chr.15: 74798973-74798973 on Build GRCh38
rs921836114 Chr.15: 74799086-74799086 on Build GRCh38
rs979119460 Chr.15: 74799080-74799080 on Build GRCh38
rs925064398 Chr.15: 74799072-74799072 on Build GRCh38
rs937878837 Chr.15: 74799096-74799097 on Build GRCh38
rs184650368 Chr.15: 74799010-74799010 on Build GRCh38
rs1007772765 Chr.15: 74799032-74799032 on Build GRCh38
rs991804815 Chr.15: 74799056-74799056 on Build GRCh38
rs1020690196 Chr.15: 74799069-74799069 on Build GRCh38
rs201168194 Chr.15: 74798959-74798959 on Build GRCh38
rs878910762 Chr.15: 74799070-74799070 on Build GRCh38
rs200650939 Chr.15: 74798965-74798965 on Build GRCh38
rs374131299 Chr.15: 74799078-74799078 on Build GRCh38
rs562760763 Chr.15: 74798907-74798907 on Build GRCh38
rs549022593 Chr.15: 74799059-74799059 on Build GRCh38
rs974549928 Chr.15: 74799081-74799081 on Build GRCh38

View Full Product Details