Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67184921-67185150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L1
Entrez Gene ID
283849
Gene Name
exocyst complex component 3 like 1
Gene Aliases
EXOC3L
UniGene
Hs.647356
Gene Chromosome Location
Chr.16: 67184379-67190204 on Build GRCh38
Transcripts
NM_178516.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061765 c.1742C>T p.T581M Chr.16: 67185143-67185143 on Build GRCh38
7418713 c.1823G>T p.C608F Chr.16: 67184984-67184984 on Build GRCh38
6657131 c.1761T>C p.A587A Chr.16: 67185046-67185046 on Build GRCh38
972322 c.1743G>T p.T581T Chr.16: 67185142-67185142 on Build GRCh38
3771979 c.1792C>T p.L598L Chr.16: 67185015-67185015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143965297 Chr.16: 67184962-67184962 on Build GRCh38
rs532889499 Chr.16: 67185088-67185088 on Build GRCh38
rs1025413712 Chr.16: 67185094-67185094 on Build GRCh38
rs986705197 Chr.16: 67185114-67185114 on Build GRCh38
rs748834819 Chr.16: 67185125-67185125 on Build GRCh38
rs995337026 Chr.16: 67185060-67185060 on Build GRCh38
rs762132461 Chr.16: 67185052-67185052 on Build GRCh38
rs1012410742 Chr.16: 67185013-67185013 on Build GRCh38
rs748779711 Chr.16: 67185029-67185029 on Build GRCh38
rs773713271 Chr.16: 67185036-67185044 on Build GRCh38
rs773886781 Chr.16: 67185062-67185062 on Build GRCh38
rs764453867 Chr.16: 67184950-67184950 on Build GRCh38
rs746848602 Chr.16: 67184978-67184978 on Build GRCh38
rs755106466 Chr.16: 67185085-67185085 on Build GRCh38
rs750919533 Chr.16: 67184966-67184966 on Build GRCh38
rs766527114 Chr.16: 67184971-67184971 on Build GRCh38
rs369614815 Chr.16: 67184974-67184974 on Build GRCh38
rs766582125 Chr.16: 67185065-67185065 on Build GRCh38
rs111513602 Chr.16: 67185093-67185093 on Build GRCh38
rs62057953 Chr.16: 67185061-67185061 on Build GRCh38
rs745760978 Chr.16: 67185009-67185009 on Build GRCh38
rs774633963 Chr.16: 67184952-67184952 on Build GRCh38
rs757029621 Chr.16: 67185101-67185101 on Build GRCh38
rs754032003 Chr.16: 67185071-67185071 on Build GRCh38
rs774919316 Chr.16: 67185027-67185027 on Build GRCh38
rs761198789 Chr.16: 67185063-67185063 on Build GRCh38
rs903477219 Chr.16: 67184957-67184957 on Build GRCh38
rs772067952 Chr.16: 67185123-67185124 on Build GRCh38
rs762231652 Chr.16: 67184960-67184960 on Build GRCh38
rs370913149 Chr.16: 67185026-67185026 on Build GRCh38
rs752945742 Chr.16: 67185098-67185098 on Build GRCh38
rs964039669 Chr.16: 67185126-67185126 on Build GRCh38
rs756098820 Chr.16: 67185117-67185117 on Build GRCh38
rs781044046 Chr.16: 67184994-67184994 on Build GRCh38
rs201243773 Chr.16: 67185132-67185132 on Build GRCh38
rs1043403885 Chr.16: 67185000-67185000 on Build GRCh38
rs769294128 Chr.16: 67185017-67185017 on Build GRCh38
rs896125966 Chr.16: 67184980-67184980 on Build GRCh38
rs756284974 Chr.16: 67184968-67184968 on Build GRCh38
rs779184172 Chr.16: 67184977-67184977 on Build GRCh38
rs745866906 Chr.16: 67185048-67185087 on Build GRCh38
rs757048997 Chr.16: 67184990-67184990 on Build GRCh38
rs996521015 Chr.16: 67185019-67185019 on Build GRCh38
rs750370383 Chr.16: 67185102-67185102 on Build GRCh38
rs899464688 Chr.16: 67185001-67185001 on Build GRCh38
rs768257010 Chr.16: 67185030-67185030 on Build GRCh38
rs762522248 Chr.16: 67184949-67184949 on Build GRCh38
rs890861988 Chr.16: 67185032-67185032 on Build GRCh38
rs376511701 Chr.16: 67185035-67185035 on Build GRCh38

View Full Product Details