Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81932511-81932738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D4
Entrez Gene ID
389761
Gene Name
SPATA31 subfamily D member 4
Gene Aliases
FAM75D4
UniGene
Hs.722630
Gene Chromosome Location
Chr.9: 81928428-81934998 on Build GRCh38
Transcripts
NM_001145197.1

SNP Detail

SNP ID Chromosome Location
rs749829602 Chr.9: 81932670-81932670 on Build GRCh38
rs779794288 Chr.9: 81932552-81932552 on Build GRCh38
rs762965436 Chr.9: 81932584-81932584 on Build GRCh38
rs759647627 Chr.9: 81932685-81932685 on Build GRCh38
rs748577961 Chr.9: 81932573-81932573 on Build GRCh38
rs774539489 Chr.9: 81932682-81932682 on Build GRCh38
rs751119096 Chr.9: 81932648-81932648 on Build GRCh38
rs1010345281 Chr.9: 81932553-81932553 on Build GRCh38
rs544627271 Chr.9: 81932600-81932600 on Build GRCh38
rs770322750 Chr.9: 81932577-81932577 on Build GRCh38
rs765321413 Chr.9: 81932613-81932613 on Build GRCh38
rs778190586 Chr.9: 81932668-81932668 on Build GRCh38
rs750397437 Chr.9: 81932616-81932616 on Build GRCh38
rs761640564 Chr.9: 81932705-81932705 on Build GRCh38
rs577362746 Chr.9: 81932569-81932569 on Build GRCh38
rs753708435 Chr.9: 81932605-81932605 on Build GRCh38
rs771007536 Chr.9: 81932585-81932585 on Build GRCh38
rs975368042 Chr.9: 81932619-81932619 on Build GRCh38
rs558911358 Chr.9: 81932546-81932546 on Build GRCh38
rs574752306 Chr.9: 81932688-81932688 on Build GRCh38
rs767058396 Chr.9: 81932641-81932641 on Build GRCh38
rs768859637 Chr.9: 81932568-81932568 on Build GRCh38
rs750346063 Chr.9: 81932714-81932714 on Build GRCh38
rs781376500 Chr.9: 81932657-81932657 on Build GRCh38
rs1019113490 Chr.9: 81932556-81932557 on Build GRCh38
rs773270192 Chr.9: 81932578-81932578 on Build GRCh38
rs770950247 Chr.9: 81932679-81932679 on Build GRCh38
rs534281288 Chr.9: 81932538-81932538 on Build GRCh38
rs562847822 Chr.9: 81932607-81932607 on Build GRCh38
rs774400141 Chr.9: 81932587-81932587 on Build GRCh38
rs368419977 Chr.9: 81932535-81932535 on Build GRCh38
rs796937489 Chr.9: 81932534-81932534 on Build GRCh38
rs754705400 Chr.9: 81932649-81932649 on Build GRCh38
rs776274672 Chr.9: 81932696-81932696 on Build GRCh38
rs765128830 Chr.9: 81932713-81932713 on Build GRCh38
rs758108109 Chr.9: 81932550-81932550 on Build GRCh38
rs779738926 Chr.9: 81932621-81932621 on Build GRCh38
rs746677727 Chr.9: 81932558-81932558 on Build GRCh38
rs748607319 Chr.9: 81932663-81932663 on Build GRCh38
rs909620140 Chr.9: 81932707-81932707 on Build GRCh38
rs770159427 Chr.9: 81932665-81932665 on Build GRCh38

View Full Product Details