Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100623514-100623761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L3
Entrez Gene ID
283431
Gene Name
growth arrest specific 2 like 3
Gene Aliases
G2L3
UniGene
Hs.20575
Gene Chromosome Location
Chr.12: 100573661-100628288 on Build GRCh38
Transcripts
NM_001303130.1 NM_001303131.1 NM_174942.2 XM_005268804.3 XM_011538219.2 XM_011538220.2 XM_011538221.2 XM_017019206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841002 c.865G>T p.E289* Chr.12: 100623670-100623670 on Build GRCh38
4686447 c.774T>C p.H258H Chr.12: 100623579-100623579 on Build GRCh38
5890695 c.815G>A p.G272E Chr.12: 100623620-100623620 on Build GRCh38
5417510 c.842G>A p.R281Q Chr.12: 100623647-100623647 on Build GRCh38
4914368 c.895G>T p.V299F Chr.12: 100623700-100623700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767269799 Chr.12: 100623670-100623670 on Build GRCh38
rs550402512 Chr.12: 100623710-100623710 on Build GRCh38
rs1024731201 Chr.12: 100623715-100623715 on Build GRCh38
rs771077255 Chr.12: 100623647-100623647 on Build GRCh38
rs143386607 Chr.12: 100623718-100623718 on Build GRCh38
rs747309935 Chr.12: 100623609-100623609 on Build GRCh38
rs201339445 Chr.12: 100623692-100623692 on Build GRCh38
rs750479911 Chr.12: 100623595-100623595 on Build GRCh38
rs369864279 Chr.12: 100623590-100623590 on Build GRCh38
rs761698076 Chr.12: 100623659-100623660 on Build GRCh38
rs1001478044 Chr.12: 100623552-100623552 on Build GRCh38
rs969535834 Chr.12: 100623547-100623547 on Build GRCh38
rs776517616 Chr.12: 100623545-100623545 on Build GRCh38
rs745473728 Chr.12: 100623730-100623730 on Build GRCh38
rs762377847 Chr.12: 100623655-100623655 on Build GRCh38
rs529481088 Chr.12: 100623727-100623727 on Build GRCh38
rs957204912 Chr.12: 100623576-100623576 on Build GRCh38
rs760545559 Chr.12: 100623687-100623687 on Build GRCh38
rs542802698 Chr.12: 100623739-100623739 on Build GRCh38
rs778914434 Chr.12: 100623593-100623593 on Build GRCh38
rs781717358 Chr.12: 100623637-100623637 on Build GRCh38
rs765964815 Chr.12: 100623553-100623553 on Build GRCh38
rs770542445 Chr.12: 100623657-100623657 on Build GRCh38
rs749864541 Chr.12: 100623726-100623726 on Build GRCh38
rs756076096 Chr.12: 100623592-100623592 on Build GRCh38
rs769117819 Chr.12: 100623621-100623621 on Build GRCh38
rs1005749689 Chr.12: 100623682-100623682 on Build GRCh38
rs759248021 Chr.12: 100623566-100623566 on Build GRCh38
rs199712487 Chr.12: 100623629-100623629 on Build GRCh38
rs143410282 Chr.12: 100623602-100623602 on Build GRCh38
rs766524774 Chr.12: 100623693-100623693 on Build GRCh38
rs753878982 Chr.12: 100623690-100623690 on Build GRCh38
rs752609151 Chr.12: 100623591-100623591 on Build GRCh38
rs375615392 Chr.12: 100623563-100623563 on Build GRCh38
rs755279811 Chr.12: 100623699-100623699 on Build GRCh38
rs781651469 Chr.12: 100623704-100623704 on Build GRCh38
rs770449052 Chr.12: 100623646-100623646 on Build GRCh38
rs748629260 Chr.12: 100623642-100623642 on Build GRCh38
rs773862403 Chr.12: 100623665-100623665 on Build GRCh38
rs765039375 Chr.12: 100623735-100623735 on Build GRCh38
rs771723176 Chr.12: 100623738-100623738 on Build GRCh38
rs778139968 Chr.12: 100623725-100623725 on Build GRCh38
rs371651349 Chr.12: 100623673-100623673 on Build GRCh38
rs113932254 Chr.12: 100623612-100623612 on Build GRCh38
rs762619994 Chr.12: 100623551-100623551 on Build GRCh38
rs751697930 Chr.12: 100623696-100623696 on Build GRCh38
rs1469385 Chr.12: 100623667-100623667 on Build GRCh38
rs1041114917 Chr.12: 100623583-100623583 on Build GRCh38
rs764022350 Chr.12: 100623689-100623689 on Build GRCh38
rs773953129 Chr.12: 100623728-100623728 on Build GRCh38

View Full Product Details