Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54499274-54499543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCKAP1L
Entrez Gene ID
3071
Gene Name
NCK associated protein 1 like
Gene Aliases
HEM1
UniGene
Hs.182014
Gene Chromosome Location
Chr.12: 54497711-54543115 on Build GRCh38
Transcripts
NM_001184976.1 NM_005337.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043207 c.119A>T p.K40I Chr.12: 54499371-54499371 on Build GRCh38
325924 c.174C>G p.I58M Chr.12: 54499426-54499426 on Build GRCh38
78152 c.212C>T p.T71M Chr.12: 54499464-54499464 on Build GRCh38
1933965 c.203G>A p.R68Q Chr.12: 54499455-54499455 on Build GRCh38
1362800 c.213G>A p.T71T Chr.12: 54499465-54499465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753534065 Chr.12: 54499456-54499456 on Build GRCh38
rs757524459 Chr.12: 54499315-54499315 on Build GRCh38
rs755898531 Chr.12: 54499347-54499347 on Build GRCh38
rs534346021 Chr.12: 54499446-54499446 on Build GRCh38
rs202229208 Chr.12: 54499455-54499455 on Build GRCh38
rs781373740 Chr.12: 54499325-54499325 on Build GRCh38
rs762455884 Chr.12: 54499426-54499428 on Build GRCh38
rs771699995 Chr.12: 54499490-54499490 on Build GRCh38
rs761772595 Chr.12: 54499443-54499443 on Build GRCh38
rs77427130 Chr.12: 54499448-54499448 on Build GRCh38
rs751556153 Chr.12: 54499312-54499312 on Build GRCh38
rs939057142 Chr.12: 54499335-54499335 on Build GRCh38
rs749263836 Chr.12: 54499359-54499359 on Build GRCh38
rs780024854 Chr.12: 54499351-54499351 on Build GRCh38
rs777622993 Chr.12: 54499515-54499515 on Build GRCh38
rs770267065 Chr.12: 54499391-54499391 on Build GRCh38
rs754963828 Chr.12: 54499460-54499460 on Build GRCh38
rs777242707 Chr.12: 54499418-54499418 on Build GRCh38
rs751713700 Chr.12: 54499441-54499441 on Build GRCh38
rs747561683 Chr.12: 54499370-54499370 on Build GRCh38
rs201460980 Chr.12: 54499410-54499410 on Build GRCh38
rs267603545 Chr.12: 54499426-54499426 on Build GRCh38
rs778514695 Chr.12: 54499362-54499362 on Build GRCh38
rs1057499805 Chr.12: 54499397-54499397 on Build GRCh38
rs772964218 Chr.12: 54499390-54499390 on Build GRCh38
rs1013471871 Chr.12: 54499423-54499423 on Build GRCh38
rs368060986 Chr.12: 54499334-54499334 on Build GRCh38
rs775656798 Chr.12: 54499409-54499409 on Build GRCh38
rs537538241 Chr.12: 54499464-54499464 on Build GRCh38
rs771900121 Chr.12: 54499374-54499374 on Build GRCh38
rs371786749 Chr.12: 54499336-54499336 on Build GRCh38
rs778692151 Chr.12: 54499465-54499465 on Build GRCh38
rs747734129 Chr.12: 54499468-54499468 on Build GRCh38
rs148094880 Chr.12: 54499424-54499424 on Build GRCh38

View Full Product Details