Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 56815552-56815789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K1
Entrez Gene ID
4214
Gene Name
mitogen-activated protein kinase kinase kinase 1
Gene Aliases
MAPKKK1, MEKK, MEKK 1, MEKK1, SRXY6
UniGene
Hs.653654
Gene Chromosome Location
Chr.5: 56815073-56896152 on Build GRCh38
Transcripts
NM_005921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6914272 c.179G>A p.R60Q Chr.5: 56815752-56815752 on Build GRCh38
6986069 c.2_3insGGC p.A5_G6insA Chr.5: 56815575-56815576 on Build GRCh38
3393615 c.24C>T p.R8R Chr.5: 56815597-56815597 on Build GRCh38
6927102 c.50C>T p.A17V Chr.5: 56815623-56815623 on Build GRCh38
5752869 c.21_22insC p.R8fs*16 Chr.5: 56815594-56815595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758267902 Chr.5: 56815633-56815633 on Build GRCh38
rs980247055 Chr.5: 56815663-56815663 on Build GRCh38
rs1051867310 Chr.5: 56815711-56815711 on Build GRCh38
rs757493757 Chr.5: 56815656-56815656 on Build GRCh38
rs950847304 Chr.5: 56815649-56815649 on Build GRCh38
rs1053641526 Chr.5: 56815678-56815678 on Build GRCh38
rs994813732 Chr.5: 56815627-56815627 on Build GRCh38
rs1027679613 Chr.5: 56815647-56815647 on Build GRCh38
rs577708359 Chr.5: 56815749-56815749 on Build GRCh38
rs919980796 Chr.5: 56815628-56815628 on Build GRCh38
rs544674438 Chr.5: 56815760-56815760 on Build GRCh38
rs971549264 Chr.5: 56815662-56815662 on Build GRCh38
rs892176374 Chr.5: 56815697-56815697 on Build GRCh38
rs891002481 Chr.5: 56815604-56815604 on Build GRCh38
rs765282250 Chr.5: 56815617-56815617 on Build GRCh38
rs974164827 Chr.5: 56815626-56815626 on Build GRCh38
rs189140884 Chr.5: 56815738-56815738 on Build GRCh38
rs185050655 Chr.5: 56815654-56815654 on Build GRCh38
rs886480221 Chr.5: 56815770-56815770 on Build GRCh38
rs777667605 Chr.5: 56815637-56815637 on Build GRCh38
rs745954108 Chr.5: 56815716-56815716 on Build GRCh38
rs983153631 Chr.5: 56815635-56815635 on Build GRCh38
rs1039577956 Chr.5: 56815739-56815739 on Build GRCh38
rs936484973 Chr.5: 56815664-56815664 on Build GRCh38
rs951507666 Chr.5: 56815629-56815629 on Build GRCh38
rs779149827 Chr.5: 56815575-56815576 on Build GRCh38
rs1050828981 Chr.5: 56815734-56815734 on Build GRCh38
rs983587660 Chr.5: 56815652-56815652 on Build GRCh38
rs555957417 Chr.5: 56815609-56815609 on Build GRCh38
rs749840532 Chr.5: 56815748-56815748 on Build GRCh38
rs776502274 Chr.5: 56815593-56815593 on Build GRCh38
rs1008473777 Chr.5: 56815719-56815719 on Build GRCh38
rs927520159 Chr.5: 56815680-56815680 on Build GRCh38
rs1009619802 Chr.5: 56815584-56815584 on Build GRCh38
rs769021328 Chr.5: 56815763-56815763 on Build GRCh38
rs759046760 Chr.5: 56815588-56815588 on Build GRCh38
rs1018559368 Chr.5: 56815725-56815725 on Build GRCh38
rs949301616 Chr.5: 56815638-56815638 on Build GRCh38
rs868220086 Chr.5: 56815595-56815595 on Build GRCh38
rs192120973 Chr.5: 56815618-56815618 on Build GRCh38
rs746070735 Chr.5: 56815742-56815743 on Build GRCh38
rs781353360 Chr.5: 56815672-56815672 on Build GRCh38
rs1009323622 Chr.5: 56815707-56815707 on Build GRCh38
rs769918068 Chr.5: 56815736-56815736 on Build GRCh38
rs890879399 Chr.5: 56815737-56815737 on Build GRCh38

View Full Product Details