Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45094508-45094775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679668 c.4396-3_4396-2insC p.? Chr.15: 45094693-45094694 on Build GRCh38
5515560 c.4494C>A p.F1498L Chr.15: 45094593-45094593 on Build GRCh38
3501612 c.4416C>T p.F1472F Chr.15: 45094671-45094671 on Build GRCh38
7422181 c.4478C>G p.P1493R Chr.15: 45094609-45094609 on Build GRCh38
7096356 c.4494C>G p.F1498L Chr.15: 45094593-45094593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767845393 Chr.15: 45094556-45094556 on Build GRCh38
rs777339722 Chr.15: 45094685-45094685 on Build GRCh38
rs960545052 Chr.15: 45094696-45094696 on Build GRCh38
rs764419796 Chr.15: 45094723-45094723 on Build GRCh38
rs748234399 Chr.15: 45094703-45094703 on Build GRCh38
rs144543420 Chr.15: 45094612-45094612 on Build GRCh38
rs752847404 Chr.15: 45094585-45094585 on Build GRCh38
rs576041718 Chr.15: 45094606-45094606 on Build GRCh38
rs756267461 Chr.15: 45094601-45094601 on Build GRCh38
rs770412777 Chr.15: 45094691-45094691 on Build GRCh38
rs879026158 Chr.15: 45094567-45094567 on Build GRCh38
rs754892057 Chr.15: 45094738-45094738 on Build GRCh38
rs772727433 Chr.15: 45094624-45094624 on Build GRCh38
rs372384451 Chr.15: 45094726-45094726 on Build GRCh38
rs56323146 Chr.15: 45094608-45094608 on Build GRCh38
rs761064126 Chr.15: 45094716-45094716 on Build GRCh38
rs776750341 Chr.15: 45094693-45094694 on Build GRCh38
rs368004695 Chr.15: 45094634-45094634 on Build GRCh38
rs754112119 Chr.15: 45094725-45094725 on Build GRCh38
rs751933939 Chr.15: 45094540-45094540 on Build GRCh38
rs978076544 Chr.15: 45094752-45094752 on Build GRCh38
rs774970795 Chr.15: 45094695-45094695 on Build GRCh38
rs898561291 Chr.15: 45094633-45094633 on Build GRCh38
rs1040212550 Chr.15: 45094616-45094616 on Build GRCh38
rs750546082 Chr.15: 45094642-45094642 on Build GRCh38
rs138103904 Chr.15: 45094678-45094678 on Build GRCh38
rs767753281 Chr.15: 45094648-45094648 on Build GRCh38
rs182797527 Chr.15: 45094730-45094730 on Build GRCh38
rs371970228 Chr.15: 45094655-45094655 on Build GRCh38
rs759931529 Chr.15: 45094698-45094698 on Build GRCh38
rs1023831110 Chr.15: 45094741-45094741 on Build GRCh38
rs149535976 Chr.15: 45094683-45094683 on Build GRCh38
rs766548595 Chr.15: 45094548-45094548 on Build GRCh38
rs997835124 Chr.15: 45094543-45094543 on Build GRCh38
rs376623263 Chr.15: 45094682-45094682 on Build GRCh38
rs772041683 Chr.15: 45094571-45094571 on Build GRCh38
rs575836804 Chr.15: 45094745-45094745 on Build GRCh38
rs755230491 Chr.15: 45094554-45094554 on Build GRCh38
rs768334494 Chr.15: 45094609-45094609 on Build GRCh38
rs771562080 Chr.15: 45094575-45094575 on Build GRCh38
rs556008835 Chr.15: 45094602-45094602 on Build GRCh38
rs143154243 Chr.15: 45094684-45094684 on Build GRCh38
rs748863515 Chr.15: 45094689-45094689 on Build GRCh38
rs771432635 Chr.15: 45094635-45094635 on Build GRCh38
rs376718345 Chr.15: 45094742-45094742 on Build GRCh38
rs774714033 Chr.15: 45094641-45094641 on Build GRCh38
rs546148981 Chr.15: 45094746-45094746 on Build GRCh38
rs747720952 Chr.15: 45094613-45094613 on Build GRCh38
rs759342749 Chr.15: 45094654-45094654 on Build GRCh38
rs578014563 Chr.15: 45094561-45094562 on Build GRCh38
rs766819778 Chr.15: 45094527-45094527 on Build GRCh38
rs923636133 Chr.15: 45094754-45094754 on Build GRCh38
rs1016673681 Chr.15: 45094697-45094697 on Build GRCh38
rs750472682 Chr.15: 45094731-45094731 on Build GRCh38
rs764364381 Chr.15: 45094667-45094667 on Build GRCh38
rs200785525 Chr.15: 45094679-45094679 on Build GRCh38

View Full Product Details