Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18399934-18400185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT2
Entrez Gene ID
10
Gene Name
N-acetyltransferase 2
Gene Aliases
AAC2, NAT-2, PNAT
UniGene
Hs.2
Gene Chromosome Location
Chr.8: 18386585-18401219 on Build GRCh38
Transcripts
NM_000015.2 XM_017012938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1699775 c.4G>A p.D2N Chr.8: 18400007-18400007 on Build GRCh38
5898356 c.106C>A p.P36T Chr.8: 18400109-18400109 on Build GRCh38
1643553 c.159G>T p.E53D Chr.8: 18400162-18400162 on Build GRCh38
3646975 c.63G>A p.L21L Chr.8: 18400066-18400066 on Build GRCh38
3646974 c.26G>A p.R9K Chr.8: 18400029-18400029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370796160 Chr.8: 18400132-18400132 on Build GRCh38
rs770754029 Chr.8: 18399963-18399963 on Build GRCh38
rs779874333 Chr.8: 18400141-18400141 on Build GRCh38
rs764262450 Chr.8: 18400000-18400000 on Build GRCh38
rs776798810 Chr.8: 18400072-18400072 on Build GRCh38
rs72466457 Chr.8: 18400155-18400155 on Build GRCh38
rs745585836 Chr.8: 18400051-18400051 on Build GRCh38
rs777235619 Chr.8: 18400042-18400042 on Build GRCh38
rs973955658 Chr.8: 18399968-18399968 on Build GRCh38
rs753310036 Chr.8: 18400113-18400113 on Build GRCh38
rs747536177 Chr.8: 18400035-18400035 on Build GRCh38
rs771801023 Chr.8: 18400137-18400137 on Build GRCh38
rs765487420 Chr.8: 18400082-18400082 on Build GRCh38
rs753823712 Chr.8: 18400136-18400136 on Build GRCh38
rs760627448 Chr.8: 18400065-18400065 on Build GRCh38
rs984174803 Chr.8: 18400063-18400063 on Build GRCh38
rs750811489 Chr.8: 18400099-18400099 on Build GRCh38
rs775647826 Chr.8: 18399997-18399997 on Build GRCh38
rs763283305 Chr.8: 18400100-18400100 on Build GRCh38
rs767443003 Chr.8: 18399983-18399983 on Build GRCh38
rs774237368 Chr.8: 18400045-18400045 on Build GRCh38
rs767322047 Chr.8: 18399996-18399996 on Build GRCh38
rs774110362 Chr.8: 18399978-18399978 on Build GRCh38
rs754150699 Chr.8: 18400008-18400008 on Build GRCh38
rs934741595 Chr.8: 18400058-18400058 on Build GRCh38
rs772909121 Chr.8: 18399958-18399958 on Build GRCh38
rs770525419 Chr.8: 18400043-18400043 on Build GRCh38
rs138592670 Chr.8: 18400101-18400101 on Build GRCh38
rs377338395 Chr.8: 18400109-18400109 on Build GRCh38
rs928522570 Chr.8: 18400050-18400050 on Build GRCh38
rs760616420 Chr.8: 18399999-18399999 on Build GRCh38
rs886633825 Chr.8: 18400127-18400127 on Build GRCh38
rs866596704 Chr.8: 18400139-18400139 on Build GRCh38
rs199780526 Chr.8: 18400107-18400107 on Build GRCh38
rs186884477 Chr.8: 18400011-18400011 on Build GRCh38
rs750399005 Chr.8: 18400103-18400103 on Build GRCh38
rs149283608 Chr.8: 18400124-18400124 on Build GRCh38
rs200893121 Chr.8: 18400010-18400010 on Build GRCh38
rs72466456 Chr.8: 18400032-18400032 on Build GRCh38
rs532310930 Chr.8: 18400061-18400061 on Build GRCh38
rs72554615 Chr.8: 18400114-18400114 on Build GRCh38
rs752047953 Chr.8: 18400106-18400106 on Build GRCh38
rs755689546 Chr.8: 18400038-18400038 on Build GRCh38
rs867460835 Chr.8: 18400104-18400104 on Build GRCh38
rs111750824 Chr.8: 18400151-18400151 on Build GRCh38
rs759544825 Chr.8: 18399993-18399993 on Build GRCh38
rs764315394 Chr.8: 18400071-18400071 on Build GRCh38
rs201339185 Chr.8: 18400054-18400054 on Build GRCh38
rs45477599 Chr.8: 18400073-18400073 on Build GRCh38

View Full Product Details