Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133531961-133532234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2E1
Entrez Gene ID
1571
Gene Name
cytochrome P450 family 2 subfamily E member 1
Gene Aliases
CPE1, CYP2E, P450-J, P450C2E
UniGene
Hs.12907
Gene Chromosome Location
Chr.10: 133527363-133539116 on Build GRCh38
Transcripts
NM_000773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929181 c.499G>A p.D167N Chr.10: 133532135-133532135 on Build GRCh38
5469383 c.557G>A p.R186H Chr.10: 133532193-133532193 on Build GRCh38
3367971 c.549C>G p.I183M Chr.10: 133532185-133532185 on Build GRCh38
280263 c.584A>C p.K195T Chr.10: 133532220-133532220 on Build GRCh38
355828 c.532A>C p.N178H Chr.10: 133532168-133532168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780885158 Chr.10: 133532122-133532122 on Build GRCh38
rs566294180 Chr.10: 133532150-133532150 on Build GRCh38
rs75795289 Chr.10: 133532134-133532134 on Build GRCh38
rs968119639 Chr.10: 133532002-133532002 on Build GRCh38
rs752959099 Chr.10: 133532204-133532204 on Build GRCh38
rs747523205 Chr.10: 133532073-133532073 on Build GRCh38
rs997336977 Chr.10: 133532023-133532023 on Build GRCh38
rs140500826 Chr.10: 133532192-133532192 on Build GRCh38
rs923179921 Chr.10: 133532157-133532157 on Build GRCh38
rs201427783 Chr.10: 133532176-133532176 on Build GRCh38
rs755208649 Chr.10: 133532142-133532142 on Build GRCh38
rs769163444 Chr.10: 133532170-133532177 on Build GRCh38
rs1022723490 Chr.10: 133531990-133531990 on Build GRCh38
rs568558058 Chr.10: 133532170-133532170 on Build GRCh38
rs79549556 Chr.10: 133532179-133532179 on Build GRCh38
rs769469905 Chr.10: 133532094-133532094 on Build GRCh38
rs749371010 Chr.10: 133532161-133532161 on Build GRCh38
rs6413419 Chr.10: 133532171-133532171 on Build GRCh38
rs56040284 Chr.10: 133532159-133532159 on Build GRCh38
rs76138620 Chr.10: 133532180-133532180 on Build GRCh38
rs546804720 Chr.10: 133532071-133532071 on Build GRCh38
rs139133362 Chr.10: 133532152-133532152 on Build GRCh38
rs919286179 Chr.10: 133532105-133532105 on Build GRCh38
rs763652251 Chr.10: 133532102-133532102 on Build GRCh38
rs776876434 Chr.10: 133532089-133532089 on Build GRCh38
rs750134672 Chr.10: 133532022-133532022 on Build GRCh38
rs374523768 Chr.10: 133532093-133532093 on Build GRCh38
rs548262477 Chr.10: 133532160-133532160 on Build GRCh38
rs60452492 Chr.10: 133532153-133532153 on Build GRCh38
rs569885410 Chr.10: 133532051-133532051 on Build GRCh38
rs756169972 Chr.10: 133532158-133532158 on Build GRCh38
rs200871390 Chr.10: 133532193-133532193 on Build GRCh38
rs750206115 Chr.10: 133532141-133532141 on Build GRCh38
rs753014113 Chr.10: 133532143-133532143 on Build GRCh38
rs958885659 Chr.10: 133532043-133532043 on Build GRCh38
rs775164673 Chr.10: 133532097-133532097 on Build GRCh38
rs771264192 Chr.10: 133532078-133532078 on Build GRCh38
rs758535504 Chr.10: 133532147-133532147 on Build GRCh38
rs201167793 Chr.10: 133532129-133532129 on Build GRCh38
rs971765663 Chr.10: 133532084-133532084 on Build GRCh38
rs527357372 Chr.10: 133532007-133532007 on Build GRCh38
rs9919386 Chr.10: 133532099-133532099 on Build GRCh38
rs557994567 Chr.10: 133532162-133532162 on Build GRCh38

View Full Product Details