Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11677546-11677773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAD2L2
Entrez Gene ID
10459
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Gene Aliases
MAD2B, POLZ2, REV7
UniGene
Hs.19400
Gene Chromosome Location
Chr.1: 11674480-11691788 on Build GRCh38
Transcripts
NM_001127325.1 NM_006341.3 XM_011540507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1626338 c.214A>C p.K72Q Chr.1: 11677560-11677560 on Build GRCh38
5518108 c.290C>T p.T97M Chr.1: 11677574-11677574 on Build GRCh38
6251553 c.200C>A p.T67K Chr.1: 11677574-11677574 on Build GRCh38
4944484 c.214A>C p.K72Q Chr.1: 11677560-11677560 on Build GRCh38
2178629 c.211G>A p.V71I Chr.1: 11677563-11677563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766344284 Chr.1: 11677632-11677632 on Build GRCh38
rs954968986 Chr.1: 11677655-11677655 on Build GRCh38
rs147937370 Chr.1: 11677572-11677572 on Build GRCh38
rs995733453 Chr.1: 11677750-11677750 on Build GRCh38
rs140482494 Chr.1: 11677600-11677600 on Build GRCh38
rs557890629 Chr.1: 11677706-11677706 on Build GRCh38
rs2233017 Chr.1: 11677710-11677710 on Build GRCh38
rs910683988 Chr.1: 11677751-11677751 on Build GRCh38
rs372156161 Chr.1: 11677637-11677637 on Build GRCh38
rs768584114 Chr.1: 11677573-11677573 on Build GRCh38
rs927560950 Chr.1: 11677747-11677747 on Build GRCh38
rs374279848 Chr.1: 11677671-11677671 on Build GRCh38
rs886844600 Chr.1: 11677604-11677604 on Build GRCh38
rs374564736 Chr.1: 11677665-11677665 on Build GRCh38
rs555043921 Chr.1: 11677719-11677719 on Build GRCh38
rs202238543 Chr.1: 11677638-11677638 on Build GRCh38
rs979615989 Chr.1: 11677711-11677711 on Build GRCh38
rs568513319 Chr.1: 11677633-11677633 on Build GRCh38
rs188897145 Chr.1: 11677675-11677675 on Build GRCh38
rs760533952 Chr.1: 11677630-11677630 on Build GRCh38
rs373270586 Chr.1: 11677564-11677564 on Build GRCh38
rs377687352 Chr.1: 11677574-11677574 on Build GRCh38
rs767807551 Chr.1: 11677601-11677601 on Build GRCh38
rs539277831 Chr.1: 11677677-11677677 on Build GRCh38
rs1030377862 Chr.1: 11677653-11677653 on Build GRCh38
rs773571957 Chr.1: 11677603-11677603 on Build GRCh38

View Full Product Details