Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 58660026-58660251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYSM1
Entrez Gene ID
114803
Gene Name
Myb like, SWIRM and MPN domains 1
Gene Aliases
2A-DUB, 2ADUB
UniGene
Hs.744921
Gene Chromosome Location
Chr.1: 58654739-58700091 on Build GRCh38
Transcripts
NM_001085487.2 XM_006710314.3 XM_011540573.2 XM_011540574.2 XM_017000195.1 XM_017000196.1 XM_017000197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1160934 c.2346G>A p.R782R Chr.1: 58660138-58660138 on Build GRCh38
7378753 c.2404G>C p.E802Q Chr.1: 58660080-58660080 on Build GRCh38
1742418 c.2455G>A p.E819K Chr.1: 58660029-58660029 on Build GRCh38
5592036 c.2415C>T p.F805F Chr.1: 58660069-58660069 on Build GRCh38
124647 c.2371T>G p.C791G Chr.1: 58660113-58660113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760211582 Chr.1: 58660053-58660053 on Build GRCh38
rs745946472 Chr.1: 58660182-58660182 on Build GRCh38
rs767007294 Chr.1: 58660072-58660072 on Build GRCh38
rs536481394 Chr.1: 58660115-58660115 on Build GRCh38
rs866312975 Chr.1: 58660052-58660052 on Build GRCh38
rs964181993 Chr.1: 58660111-58660111 on Build GRCh38
rs992452466 Chr.1: 58660088-58660088 on Build GRCh38
rs12092264 Chr.1: 58660222-58660222 on Build GRCh38
rs763775941 Chr.1: 58660147-58660147 on Build GRCh38
rs12082192 Chr.1: 58660227-58660227 on Build GRCh38
rs201158761 Chr.1: 58660171-58660171 on Build GRCh38
rs933429406 Chr.1: 58660229-58660229 on Build GRCh38
rs780763796 Chr.1: 58660195-58660195 on Build GRCh38
rs776144063 Chr.1: 58660061-58660061 on Build GRCh38
rs757453885 Chr.1: 58660166-58660166 on Build GRCh38
rs925753698 Chr.1: 58660196-58660196 on Build GRCh38
rs765168255 Chr.1: 58660100-58660100 on Build GRCh38
rs375003312 Chr.1: 58660120-58660120 on Build GRCh38
rs376915774 Chr.1: 58660185-58660185 on Build GRCh38
rs754867617 Chr.1: 58660197-58660197 on Build GRCh38
rs1027288768 Chr.1: 58660116-58660116 on Build GRCh38
rs12239731 Chr.1: 58660162-58660162 on Build GRCh38
rs377718641 Chr.1: 58660214-58660214 on Build GRCh38
rs765974511 Chr.1: 58660057-58660057 on Build GRCh38
rs758991975 Chr.1: 58660098-58660098 on Build GRCh38

View Full Product Details