Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14181677-14181938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPI
Entrez Gene ID
149998
Gene Name
lipase I
Gene Aliases
CT17, LPDL, PLA1C, PRED5, mPA-PLA1 beta
UniGene
Hs.139907
Gene Chromosome Location
Chr.21: 14108813-14210953 on Build GRCh38
Transcripts
NM_001302998.1 NM_001302999.1 NM_001303000.1 NM_001303001.1 NM_198996.3 XM_006723965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3707984 c.652A>T p.R218* Chr.21: 14181812-14181812 on Build GRCh38
1327472 c.680T>G p.V227G Chr.21: 14181784-14181784 on Build GRCh38
7139947 c.701C>G p.S234C Chr.21: 14181763-14181763 on Build GRCh38
6161160 c.640C>T p.P214S Chr.21: 14181824-14181824 on Build GRCh38
5681940 c.682G>T p.D228Y Chr.21: 14181782-14181782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538729927 Chr.21: 14181848-14181848 on Build GRCh38
rs553068737 Chr.21: 14181844-14181844 on Build GRCh38
rs200407550 Chr.21: 14181811-14181811 on Build GRCh38
rs373206243 Chr.21: 14181747-14181747 on Build GRCh38
rs200824427 Chr.21: 14181748-14181748 on Build GRCh38
rs368826576 Chr.21: 14181798-14181798 on Build GRCh38
rs774844088 Chr.21: 14181714-14181714 on Build GRCh38
rs751340581 Chr.21: 14181842-14181842 on Build GRCh38
rs746391309 Chr.21: 14181859-14181859 on Build GRCh38
rs779781375 Chr.21: 14181778-14181778 on Build GRCh38
rs539855459 Chr.21: 14181836-14181836 on Build GRCh38
rs758262537 Chr.21: 14181841-14181841 on Build GRCh38
rs779069231 Chr.21: 14181852-14181852 on Build GRCh38
rs569704730 Chr.21: 14181809-14181809 on Build GRCh38
rs771507325 Chr.21: 14181800-14181800 on Build GRCh38
rs997868587 Chr.21: 14181815-14181815 on Build GRCh38
rs367985340 Chr.21: 14181899-14181899 on Build GRCh38
rs776066059 Chr.21: 14181869-14181869 on Build GRCh38
rs938699362 Chr.21: 14181705-14181705 on Build GRCh38
rs534877223 Chr.21: 14181873-14181873 on Build GRCh38
rs778687780 Chr.21: 14181750-14181750 on Build GRCh38
rs370914105 Chr.21: 14181759-14181759 on Build GRCh38
rs774697754 Chr.21: 14181805-14181805 on Build GRCh38
rs777826260 Chr.21: 14181843-14181843 on Build GRCh38
rs868686710 Chr.21: 14181850-14181850 on Build GRCh38
rs202244578 Chr.21: 14181716-14181716 on Build GRCh38
rs375047826 Chr.21: 14181728-14181728 on Build GRCh38
rs2822438 Chr.21: 14181709-14181709 on Build GRCh38
rs768407499 Chr.21: 14181875-14181875 on Build GRCh38
rs763651191 Chr.21: 14181823-14181823 on Build GRCh38
rs554808871 Chr.21: 14181883-14181883 on Build GRCh38
rs758215638 Chr.21: 14181760-14181760 on Build GRCh38
rs578154767 Chr.21: 14181794-14181794 on Build GRCh38
rs750388491 Chr.21: 14181838-14181838 on Build GRCh38
rs374527200 Chr.21: 14181741-14181741 on Build GRCh38
rs377507652 Chr.21: 14181879-14181879 on Build GRCh38
rs574658597 Chr.21: 14181887-14181887 on Build GRCh38
rs772694106 Chr.21: 14181862-14181862 on Build GRCh38
rs767327203 Chr.21: 14181720-14181720 on Build GRCh38
rs200367893 Chr.21: 14181895-14181895 on Build GRCh38
rs570772306 Chr.21: 14181799-14181799 on Build GRCh38
rs1016840329 Chr.21: 14181845-14181845 on Build GRCh38
rs267606078 Chr.21: 14181773-14181773 on Build GRCh38
rs963514070 Chr.21: 14181860-14181860 on Build GRCh38
rs748370686 Chr.21: 14181785-14181785 on Build GRCh38
rs377625363 Chr.21: 14181745-14181745 on Build GRCh38
rs753430316 Chr.21: 14181833-14181833 on Build GRCh38
rs760462760 Chr.21: 14181814-14181814 on Build GRCh38

View Full Product Details