Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123165289-123165514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPHOSPH9
Entrez Gene ID
10198
Gene Name
M-phase phosphoprotein 9
Gene Aliases
MPP-9, MPP9
UniGene
Hs.577404
Gene Chromosome Location
Chr.12: 123152324-123244014 on Build GRCh38
Transcripts
NM_022782.3 NR_103517.1 XM_011537740.2 XM_011537741.2 XM_017018673.1 XM_017018674.1 XM_017018675.1 XM_017018676.1 XM_017018677.1 XM_017018678.1 XM_017018679.1 XM_017018680.1 XM_017018681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2225703 c.2190G>A p.L730L Chr.12: 123165423-123165423 on Build GRCh38
936398 c.2236G>T p.A746S Chr.12: 123165377-123165377 on Build GRCh38
4204398 c.2222C>T p.T741M Chr.12: 123165391-123165391 on Build GRCh38
1177504 c.2311+3_2311+6delGAGT p.? Chr.12: 123165296-123165299 on Build GRCh38
4204399 c.2190G>A p.L730L Chr.12: 123165423-123165423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750784465 Chr.12: 123165445-123165445 on Build GRCh38
rs765860653 Chr.12: 123165365-123165365 on Build GRCh38
rs772851603 Chr.12: 123165360-123165360 on Build GRCh38
rs148807341 Chr.12: 123165344-123165344 on Build GRCh38
rs369889526 Chr.12: 123165357-123165357 on Build GRCh38
rs756674882 Chr.12: 123165456-123165456 on Build GRCh38
rs746807574 Chr.12: 123165377-123165377 on Build GRCh38
rs1716166 Chr.12: 123165354-123165354 on Build GRCh38
rs759585307 Chr.12: 123165388-123165388 on Build GRCh38
rs371715200 Chr.12: 123165335-123165335 on Build GRCh38
rs751756717 Chr.12: 123165411-123165411 on Build GRCh38
rs775448176 Chr.12: 123165395-123165395 on Build GRCh38
rs757602144 Chr.12: 123165317-123165317 on Build GRCh38
rs374291651 Chr.12: 123165387-123165387 on Build GRCh38
rs1037372626 Chr.12: 123165320-123165320 on Build GRCh38
rs377652877 Chr.12: 123165408-123165408 on Build GRCh38
rs778413906 Chr.12: 123165322-123165325 on Build GRCh38
rs767913781 Chr.12: 123165444-123165444 on Build GRCh38
rs747612987 Chr.12: 123165473-123165473 on Build GRCh38
rs746427051 Chr.12: 123165339-123165339 on Build GRCh38
rs375735988 Chr.12: 123165347-123165347 on Build GRCh38
rs770800594 Chr.12: 123165383-123165383 on Build GRCh38
rs377560994 Chr.12: 123165355-123165355 on Build GRCh38
rs763025130 Chr.12: 123165405-123165405 on Build GRCh38
rs890095009 Chr.12: 123165431-123165431 on Build GRCh38
rs575678982 Chr.12: 123165442-123165442 on Build GRCh38
rs201725214 Chr.12: 123165472-123165472 on Build GRCh38
rs756908528 Chr.12: 123165320-123165323 on Build GRCh38
rs765259820 Chr.12: 123165391-123165391 on Build GRCh38
rs199894915 Chr.12: 123165476-123165476 on Build GRCh38
rs773329281 Chr.12: 123165424-123165424 on Build GRCh38

View Full Product Details