Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75008976-75009243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266771 c.946G>T p.V316F Chr.14: 75009078-75009078 on Build GRCh38
79779 c.1017G>A p.L339L Chr.14: 75009149-75009149 on Build GRCh38
6141257 c.1012C>G p.R338G Chr.14: 75009144-75009144 on Build GRCh38
5848545 c.946G>A p.V316I Chr.14: 75009078-75009078 on Build GRCh38
699379 c.1039G>C p.D347H Chr.14: 75009171-75009171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749059127 Chr.14: 75009033-75009033 on Build GRCh38
rs773328951 Chr.14: 75009193-75009193 on Build GRCh38
rs754711911 Chr.14: 75009143-75009143 on Build GRCh38
rs1036228873 Chr.14: 75009016-75009016 on Build GRCh38
rs370756093 Chr.14: 75009199-75009199 on Build GRCh38
rs776033447 Chr.14: 75009213-75009213 on Build GRCh38
rs765712931 Chr.14: 75009105-75009105 on Build GRCh38
rs759914845 Chr.14: 75009092-75009092 on Build GRCh38
rs138817058 Chr.14: 75009059-75009059 on Build GRCh38
rs145117455 Chr.14: 75009114-75009114 on Build GRCh38
rs767183422 Chr.14: 75009062-75009062 on Build GRCh38
rs113994018 Chr.14: 75009042-75009042 on Build GRCh38
rs770275347 Chr.14: 75009201-75009201 on Build GRCh38
rs373439468 Chr.14: 75009190-75009190 on Build GRCh38
rs761530281 Chr.14: 75009214-75009214 on Build GRCh38
rs745792330 Chr.14: 75009006-75009006 on Build GRCh38
rs759031496 Chr.14: 75009175-75009177 on Build GRCh38
rs140707986 Chr.14: 75009140-75009140 on Build GRCh38
rs147596765 Chr.14: 75009131-75009131 on Build GRCh38
rs756121992 Chr.14: 75009018-75009018 on Build GRCh38
rs779998988 Chr.14: 75009137-75009137 on Build GRCh38
rs112087431 Chr.14: 75009191-75009191 on Build GRCh38
rs770918340 Chr.14: 75009198-75009198 on Build GRCh38
rs375585987 Chr.14: 75009116-75009116 on Build GRCh38
rs200849948 Chr.14: 75009112-75009112 on Build GRCh38
rs768467979 Chr.14: 75009053-75009053 on Build GRCh38
rs766123244 Chr.14: 75009096-75009098 on Build GRCh38
rs762563184 Chr.14: 75009043-75009045 on Build GRCh38
rs773407374 Chr.14: 75009077-75009077 on Build GRCh38
rs78900727 Chr.14: 75009020-75009020 on Build GRCh38
rs772125418 Chr.14: 75009074-75009074 on Build GRCh38
rs113994020 Chr.14: 75009118-75009118 on Build GRCh38
rs1046143672 Chr.14: 75009167-75009167 on Build GRCh38
rs372548739 Chr.14: 75009054-75009054 on Build GRCh38
rs104894426 Chr.14: 75009079-75009079 on Build GRCh38
rs888884716 Chr.14: 75009080-75009080 on Build GRCh38
rs904892675 Chr.14: 75009182-75009182 on Build GRCh38
rs761055346 Chr.14: 75009078-75009078 on Build GRCh38
rs778740286 Chr.14: 75009184-75009184 on Build GRCh38
rs886050766 Chr.14: 75009177-75009179 on Build GRCh38
rs747070752 Chr.14: 75009194-75009194 on Build GRCh38
rs577844198 Chr.14: 75009111-75009111 on Build GRCh38
rs777036076 Chr.14: 75009086-75009086 on Build GRCh38
rs773725419 Chr.14: 75009173-75009174 on Build GRCh38
rs749995021 Chr.14: 75009124-75009124 on Build GRCh38

View Full Product Details