Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1182869-1183142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL10
Entrez Gene ID
254173
Gene Name
tubulin tyrosine ligase like 10
Gene Aliases
TTLL5
UniGene
Hs.454835
Gene Chromosome Location
Chr.1: 1173709-1197936 on Build GRCh38
Transcripts
NM_001130045.1 NM_153254.2 XM_005244738.1 XM_011541177.2 XM_017000906.1 XM_017000907.1 XM_017000908.1 XM_017000909.1 XM_017000910.1 XM_017000911.1 XM_017000912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527474 c.989A>C p.E330A Chr.1: 1182948-1182948 on Build GRCh38
3471172 c.836C>T p.P279L Chr.1: 1183014-1183014 on Build GRCh38
4934867 c.949T>C p.S317P Chr.1: 1182908-1182908 on Build GRCh38
4737301 c.1032C>T p.D344D Chr.1: 1182991-1182991 on Build GRCh38
5627158 c.927A>G p.I309M Chr.1: 1182886-1182886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772812931 Chr.1: 1183000-1183002 on Build GRCh38
rs750448128 Chr.1: 1182978-1182978 on Build GRCh38
rs769137130 Chr.1: 1183057-1183057 on Build GRCh38
rs138091862 Chr.1: 1182985-1182985 on Build GRCh38
rs866630539 Chr.1: 1182957-1182957 on Build GRCh38
rs773618340 Chr.1: 1183074-1183074 on Build GRCh38
rs772397027 Chr.1: 1183066-1183066 on Build GRCh38
rs140929937 Chr.1: 1182950-1182950 on Build GRCh38
rs772486594 Chr.1: 1183023-1183023 on Build GRCh38
rs992410529 Chr.1: 1182981-1182981 on Build GRCh38
rs752651634 Chr.1: 1183006-1183006 on Build GRCh38
rs546816658 Chr.1: 1182939-1182939 on Build GRCh38
rs750519272 Chr.1: 1182897-1182897 on Build GRCh38
rs989567062 Chr.1: 1183067-1183067 on Build GRCh38
rs776052443 Chr.1: 1182959-1182959 on Build GRCh38
rs770731314 Chr.1: 1182938-1182938 on Build GRCh38
rs61733845 Chr.1: 1182895-1182895 on Build GRCh38
rs149500516 Chr.1: 1183034-1183034 on Build GRCh38
rs906807331 Chr.1: 1182991-1182991 on Build GRCh38
rs773422212 Chr.1: 1183024-1183024 on Build GRCh38
rs1053930094 Chr.1: 1182914-1182914 on Build GRCh38
rs930504944 Chr.1: 1183022-1183022 on Build GRCh38
rs552655548 Chr.1: 1183124-1183124 on Build GRCh38
rs748096585 Chr.1: 1182910-1182910 on Build GRCh38
rs114587574 Chr.1: 1183035-1183035 on Build GRCh38
rs112838421 Chr.1: 1183111-1183111 on Build GRCh38
rs748777928 Chr.1: 1183054-1183054 on Build GRCh38
rs532600952 Chr.1: 1183019-1183019 on Build GRCh38
rs774624126 Chr.1: 1182967-1182967 on Build GRCh38
rs776709121 Chr.1: 1183085-1183085 on Build GRCh38
rs748724530 Chr.1: 1183065-1183065 on Build GRCh38
rs141341297 Chr.1: 1182977-1182977 on Build GRCh38
rs368616691 Chr.1: 1182906-1182906 on Build GRCh38
rs779278930 Chr.1: 1183059-1183059 on Build GRCh38
rs981370190 Chr.1: 1183110-1183110 on Build GRCh38
rs777472636 Chr.1: 1182924-1182924 on Build GRCh38
rs781004642 Chr.1: 1183012-1183012 on Build GRCh38
rs757104331 Chr.1: 1183043-1183043 on Build GRCh38
rs770367062 Chr.1: 1182958-1182958 on Build GRCh38
rs771578163 Chr.1: 1182984-1182985 on Build GRCh38
rs769064031 Chr.1: 1183016-1183016 on Build GRCh38
rs375676753 Chr.1: 1182898-1182898 on Build GRCh38
rs186578890 Chr.1: 1183020-1183020 on Build GRCh38
rs747186229 Chr.1: 1183080-1183080 on Build GRCh38
rs763435422 Chr.1: 1182960-1182960 on Build GRCh38
rs753887558 Chr.1: 1183033-1183033 on Build GRCh38
rs998868797 Chr.1: 1183061-1183061 on Build GRCh38
rs777577506 Chr.1: 1183007-1183007 on Build GRCh38
rs200505408 Chr.1: 1183095-1183095 on Build GRCh38
rs376372471 Chr.1: 1182996-1182996 on Build GRCh38
rs371677298 Chr.1: 1182925-1182925 on Build GRCh38
rs779469524 Chr.1: 1183014-1183014 on Build GRCh38
rs572607586 Chr.1: 1183126-1183126 on Build GRCh38
rs939618520 Chr.1: 1183117-1183117 on Build GRCh38
rs146443945 Chr.1: 1182923-1182923 on Build GRCh38
rs71628944 Chr.1: 1183045-1183045 on Build GRCh38
rs746653227 Chr.1: 1182935-1182935 on Build GRCh38
rs569439420 Chr.1: 1183032-1183032 on Build GRCh38
rs774424198 Chr.1: 1183099-1183099 on Build GRCh38
rs369266010 Chr.1: 1183037-1183037 on Build GRCh38
rs1056688904 Chr.1: 1183123-1183123 on Build GRCh38
rs371930110 Chr.1: 1183084-1183084 on Build GRCh38
rs766515271 Chr.1: 1182992-1182992 on Build GRCh38
rs770599987 Chr.1: 1183011-1183011 on Build GRCh38
rs548840586 Chr.1: 1183051-1183051 on Build GRCh38
rs201484894 Chr.1: 1183052-1183052 on Build GRCh38
rs1053169937 Chr.1: 1183073-1183073 on Build GRCh38

View Full Product Details