Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24241814-24242057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TINF2
Entrez Gene ID
26277
Gene Name
TERF1 interacting nuclear factor 2
Gene Aliases
DKCA3, TIN2
UniGene
Hs.496191
Gene Chromosome Location
Chr.14: 24239641-24242674 on Build GRCh38
Transcripts
NM_001099274.1 NM_012461.2 XM_005267529.3 XM_011536642.2 XM_017021216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174753 c.240G>A p.L80L Chr.14: 24241947-24241947 on Build GRCh38
348918 c.246C>A p.A82A Chr.14: 24241941-24241941 on Build GRCh38
7188212 c.243A>G p.K81K Chr.14: 24241944-24241944 on Build GRCh38
6521379 c.229G>T p.A77S Chr.14: 24241958-24241958 on Build GRCh38
7174754 c.240G>A p.L80L Chr.14: 24241947-24241947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370971769 Chr.14: 24241937-24241937 on Build GRCh38
rs777733175 Chr.14: 24241966-24241966 on Build GRCh38
rs199682439 Chr.14: 24241877-24241877 on Build GRCh38
rs778990034 Chr.14: 24241839-24241839 on Build GRCh38
rs754157164 Chr.14: 24241907-24241907 on Build GRCh38
rs769487457 Chr.14: 24241859-24241859 on Build GRCh38
rs372985267 Chr.14: 24241875-24241875 on Build GRCh38
rs1031448980 Chr.14: 24242028-24242028 on Build GRCh38
rs11844422 Chr.14: 24241845-24241845 on Build GRCh38
rs145115622 Chr.14: 24241947-24241947 on Build GRCh38
rs201825289 Chr.14: 24242025-24242025 on Build GRCh38
rs764402230 Chr.14: 24241894-24241894 on Build GRCh38
rs762140055 Chr.14: 24241909-24241909 on Build GRCh38
rs934174824 Chr.14: 24241837-24241837 on Build GRCh38
rs748978859 Chr.14: 24241862-24241862 on Build GRCh38
rs759185643 Chr.14: 24241872-24241872 on Build GRCh38
rs773714974 Chr.14: 24241841-24241841 on Build GRCh38
rs749135301 Chr.14: 24241977-24241977 on Build GRCh38
rs972411405 Chr.14: 24242023-24242023 on Build GRCh38
rs373544350 Chr.14: 24242027-24242027 on Build GRCh38
rs765479562 Chr.14: 24241926-24241926 on Build GRCh38
rs201651916 Chr.14: 24242001-24242001 on Build GRCh38
rs750637502 Chr.14: 24241929-24241929 on Build GRCh38
rs778766822 Chr.14: 24242019-24242019 on Build GRCh38
rs1028369015 Chr.14: 24241842-24241842 on Build GRCh38
rs376250937 Chr.14: 24241876-24241876 on Build GRCh38
rs756752365 Chr.14: 24242024-24242025 on Build GRCh38
rs770845742 Chr.14: 24242012-24242012 on Build GRCh38
rs369327266 Chr.14: 24241852-24241852 on Build GRCh38
rs534072086 Chr.14: 24241860-24241860 on Build GRCh38
rs775877509 Chr.14: 24241880-24241880 on Build GRCh38
rs770757511 Chr.14: 24241863-24241863 on Build GRCh38
rs890080239 Chr.14: 24241873-24241873 on Build GRCh38
rs761179618 Chr.14: 24241892-24241892 on Build GRCh38
rs758462532 Chr.14: 24241934-24241934 on Build GRCh38
rs756037848 Chr.14: 24241958-24241958 on Build GRCh38
rs999024658 Chr.14: 24242020-24242020 on Build GRCh38
rs745993165 Chr.14: 24241840-24241840 on Build GRCh38
rs1043122818 Chr.14: 24241936-24241936 on Build GRCh38
rs745469311 Chr.14: 24242022-24242022 on Build GRCh38
rs1016497738 Chr.14: 24241963-24241963 on Build GRCh38

View Full Product Details