Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2055341-2055582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC2
Entrez Gene ID
7249
Gene Name
tuberous sclerosis 2
Gene Aliases
LAM, PPP1R160, TSC4
UniGene
Hs.90303
Gene Chromosome Location
Chr.16: 2047804-2088720 on Build GRCh38
Transcripts
NM_000548.4 NM_001077183.2 NM_001114382.2 NM_001318827.1 NM_001318829.1 NM_001318831.1 NM_001318832.1 XM_005255529.4 XM_005255531.4 XM_011522636.2 XM_011522637.2 XM_011522638.2 XM_011522639.2 XM_011522640.2 XM_017023615.1 XM_017023616.1 XM_017023617.1 XM_017023618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6924056 c.571C>T p.L191F Chr.16: 2055491-2055491 on Build GRCh38
7349605 c.139_*del p.? Chr.16: 2050400-2070620 on Build GRCh38
6957851 c.541G>A p.V181M Chr.16: 2055461-2055461 on Build GRCh38
4748628 c.491T>G p.V164G Chr.16: 2055411-2055411 on Build GRCh38
4748630 c.570C>G p.Y190* Chr.16: 2055490-2055490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45485591 Chr.16: 2055458-2055458 on Build GRCh38
rs982250014 Chr.16: 2055462-2055462 on Build GRCh38
rs765439112 Chr.16: 2055376-2055376 on Build GRCh38
rs137854412 Chr.16: 2055395-2055396 on Build GRCh38
rs748692173 Chr.16: 2055424-2055424 on Build GRCh38
rs372028794 Chr.16: 2055370-2055370 on Build GRCh38
rs1007260605 Chr.16: 2055560-2055560 on Build GRCh38
rs757054946 Chr.16: 2055388-2055388 on Build GRCh38
rs137854172 Chr.16: 2055509-2055510 on Build GRCh38
rs137854332 Chr.16: 2055488-2055489 on Build GRCh38
rs137854235 Chr.16: 2055402-2055403 on Build GRCh38
rs45470502 Chr.16: 2055468-2055468 on Build GRCh38
rs137854359 Chr.16: 2055489-2055490 on Build GRCh38
rs397515071 Chr.16: 2055397-2055397 on Build GRCh38
rs764218079 Chr.16: 2055486-2055486 on Build GRCh38
rs771326069 Chr.16: 2055395-2055395 on Build GRCh38
rs397515120 Chr.16: 2055485-2055495 on Build GRCh38
rs533989586 Chr.16: 2055536-2055536 on Build GRCh38
rs867879148 Chr.16: 2055449-2055449 on Build GRCh38
rs137854207 Chr.16: 2055449-2055452 on Build GRCh38
rs587778733 Chr.16: 2055515-2055515 on Build GRCh38
rs747538587 Chr.16: 2055441-2055441 on Build GRCh38
rs774703938 Chr.16: 2055493-2055495 on Build GRCh38
rs773642257 Chr.16: 2055366-2055366 on Build GRCh38
rs371179005 Chr.16: 2055553-2055558 on Build GRCh38
rs137854122 Chr.16: 2055408-2055409 on Build GRCh38
rs1800720 Chr.16: 2055399-2055399 on Build GRCh38
rs941407136 Chr.16: 2055508-2055508 on Build GRCh38
rs140749654 Chr.16: 2055559-2055559 on Build GRCh38
rs200647260 Chr.16: 2055386-2055386 on Build GRCh38
rs755728007 Chr.16: 2055421-2055421 on Build GRCh38
rs746167724 Chr.16: 2055539-2055539 on Build GRCh38
rs773662047 Chr.16: 2055438-2055438 on Build GRCh38
rs780938957 Chr.16: 2055398-2055398 on Build GRCh38
rs775642056 Chr.16: 2055478-2055478 on Build GRCh38
rs774445870 Chr.16: 2055556-2055556 on Build GRCh38
rs45517115 Chr.16: 2055518-2055518 on Build GRCh38
rs45517112 Chr.16: 2055497-2055497 on Build GRCh38
rs983899092 Chr.16: 2055432-2055432 on Build GRCh38
rs760047438 Chr.16: 2055544-2055545 on Build GRCh38
rs772413684 Chr.16: 2055363-2055363 on Build GRCh38
rs777052621 Chr.16: 2055373-2055373 on Build GRCh38
rs750174170 Chr.16: 2055502-2055502 on Build GRCh38
rs954041989 Chr.16: 2055417-2055417 on Build GRCh38
rs45517110 Chr.16: 2055400-2055400 on Build GRCh38
rs45517114 Chr.16: 2055513-2055513 on Build GRCh38
rs779566253 Chr.16: 2055422-2055422 on Build GRCh38
rs369364803 Chr.16: 2055552-2055552 on Build GRCh38
rs45463996 Chr.16: 2055524-2055524 on Build GRCh38
rs397514899 Chr.16: 2055522-2055522 on Build GRCh38
rs747510748 Chr.16: 2055531-2055531 on Build GRCh38
rs775630113 Chr.16: 2055548-2055548 on Build GRCh38
rs200473689 Chr.16: 2055375-2055375 on Build GRCh38
rs1007277184 Chr.16: 2055369-2055369 on Build GRCh38
rs373300339 Chr.16: 2055554-2055554 on Build GRCh38
rs397515309 Chr.16: 2055505-2055505 on Build GRCh38
rs148325559 Chr.16: 2055503-2055503 on Build GRCh38
rs201589515 Chr.16: 2055385-2055385 on Build GRCh38
rs45484992 Chr.16: 2055521-2055521 on Build GRCh38
rs772641112 Chr.16: 2055436-2055436 on Build GRCh38
rs397515059 Chr.16: 2055493-2055493 on Build GRCh38
rs1037449903 Chr.16: 2055529-2055529 on Build GRCh38
rs730880012 Chr.16: 2055487-2055488 on Build GRCh38
rs758521946 Chr.16: 2055413-2055413 on Build GRCh38
rs777105764 Chr.16: 2055455-2055455 on Build GRCh38
rs760883303 Chr.16: 2055367-2055367 on Build GRCh38
rs45473598 Chr.16: 2055406-2055406 on Build GRCh38
rs45517113 Chr.16: 2055506-2055506 on Build GRCh38
rs377335730 Chr.16: 2055540-2055540 on Build GRCh38
rs754505860 Chr.16: 2055510-2055510 on Build GRCh38
rs771515880 Chr.16: 2055533-2055533 on Build GRCh38
rs397515229 Chr.16: 2055429-2055430 on Build GRCh38
rs45517111 Chr.16: 2055416-2055416 on Build GRCh38
rs137853991 Chr.16: 2055449-2055449 on Build GRCh38
rs397515140 Chr.16: 2055523-2055523 on Build GRCh38
rs752669481 Chr.16: 2055379-2055379 on Build GRCh38
rs1016698636 Chr.16: 2055465-2055465 on Build GRCh38
rs772569403 Chr.16: 2055552-2055557 on Build GRCh38
rs137854306 Chr.16: 2055401-2055401 on Build GRCh38
rs45481496 Chr.16: 2055402-2055402 on Build GRCh38
rs137854300 Chr.16: 2055511-2055511 on Build GRCh38
rs397515099 Chr.16: 2055490-2055490 on Build GRCh38
rs137854277 Chr.16: 2055445-2055445 on Build GRCh38
rs199991910 Chr.16: 2055472-2055472 on Build GRCh38
rs45505405 Chr.16: 2055480-2055480 on Build GRCh38
rs748724027 Chr.16: 2055362-2055362 on Build GRCh38
rs45460895 Chr.16: 2055520-2055520 on Build GRCh38
rs761704292 Chr.16: 2055496-2055496 on Build GRCh38
rs548831116 Chr.16: 2055382-2055382 on Build GRCh38

View Full Product Details