Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71571465-71571702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAT
Entrez Gene ID
6898
Gene Name
tyrosine aminotransferase
Gene Aliases
-
UniGene
Hs.161640
Gene Chromosome Location
Chr.16: 71566851-71577095 on Build GRCh38
Transcripts
NM_000353.2 NR_103851.1 NR_103852.1

Gene Symbol
TAT-AS1
Entrez Gene ID
100132529
Gene Name
TAT antisense RNA 1
Gene Aliases
-
UniGene
Hs.710296
Gene Chromosome Location
Chr.16: 71565016-71572438 on Build GRCh38
Transcripts
NM_000353.2 NR_103851.1 NR_103852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1228531 c.722G>A p.C241Y Chr.16: 71571643-71571643 on Build GRCh38
5385321 c.729C>T p.P243P Chr.16: 71571636-71571636 on Build GRCh38
6797048 c.717G>A p.R239R Chr.16: 71571648-71571648 on Build GRCh38
7309690 c.707-9delT p.? Chr.16: 71571667-71571667 on Build GRCh38
460466 c.744G>C p.E248D Chr.16: 71571621-71571621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005382525 Chr.16: 71571660-71571660 on Build GRCh38
rs937167512 Chr.16: 71571541-71571541 on Build GRCh38
rs376656472 Chr.16: 71571662-71571662 on Build GRCh38
rs767912599 Chr.16: 71571601-71571601 on Build GRCh38
rs373941496 Chr.16: 71571673-71571673 on Build GRCh38
rs886052273 Chr.16: 71571661-71571661 on Build GRCh38
rs868121643 Chr.16: 71571642-71571642 on Build GRCh38
rs201026211 Chr.16: 71571619-71571619 on Build GRCh38
rs913398154 Chr.16: 71571516-71571516 on Build GRCh38
rs895881765 Chr.16: 71571655-71571655 on Build GRCh38
rs940441245 Chr.16: 71571637-71571637 on Build GRCh38
rs755559658 Chr.16: 71571645-71571645 on Build GRCh38
rs369023700 Chr.16: 71571589-71571589 on Build GRCh38
rs752308214 Chr.16: 71571653-71571653 on Build GRCh38
rs927133189 Chr.16: 71571529-71571529 on Build GRCh38
rs779258482 Chr.16: 71571650-71571650 on Build GRCh38
rs776580828 Chr.16: 71571584-71571584 on Build GRCh38
rs770643888 Chr.16: 71571577-71571577 on Build GRCh38
rs761791218 Chr.16: 71571598-71571598 on Build GRCh38
rs746819529 Chr.16: 71571569-71571569 on Build GRCh38
rs917923676 Chr.16: 71571505-71571509 on Build GRCh38
rs761127598 Chr.16: 71571607-71571607 on Build GRCh38
rs774525419 Chr.16: 71571596-71571596 on Build GRCh38
rs750551095 Chr.16: 71571506-71571506 on Build GRCh38
rs777390818 Chr.16: 71571557-71571557 on Build GRCh38
rs754325905 Chr.16: 71571644-71571644 on Build GRCh38
rs769594175 Chr.16: 71571593-71571593 on Build GRCh38
rs971329327 Chr.16: 71571515-71571515 on Build GRCh38
rs548467053 Chr.16: 71571603-71571603 on Build GRCh38
rs765514944 Chr.16: 71571500-71571500 on Build GRCh38
rs766814264 Chr.16: 71571611-71571611 on Build GRCh38

View Full Product Details