Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21971466-21971701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD9
Entrez Gene ID
255762
Gene Name
PDZ domain containing 9
Gene Aliases
C16orf65
UniGene
Hs.98244
Gene Chromosome Location
Chr.16: 21957540-22001137 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Gene Symbol
UQCRC2
Entrez Gene ID
7385
Gene Name
ubiquinol-cytochrome c reductase core protein II
Gene Aliases
MC3DN5, QCR2, UQCR2
UniGene
Hs.528803
Gene Chromosome Location
Chr.16: 21953064-21983660 on Build GRCh38
Transcripts
XM_011545785.2 XM_017023109.1 XM_017023110.1 NM_003366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242184 c.689T>C p.L230P Chr.16: 21971543-21971543 on Build GRCh38
3817507 c.757T>C p.Y253H Chr.16: 21971611-21971611 on Build GRCh38
968516 c.742G>T p.G248C Chr.16: 21971596-21971596 on Build GRCh38
6343395 c.728G>T p.G243V Chr.16: 21971582-21971582 on Build GRCh38
6464272 c.688C>T p.L230L Chr.16: 21971542-21971542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11863893 Chr.16: 21971615-21971615 on Build GRCh38
rs771775129 Chr.16: 21971628-21971628 on Build GRCh38
rs929755174 Chr.16: 21971511-21971511 on Build GRCh38
rs986874832 Chr.16: 21971529-21971530 on Build GRCh38
rs376000972 Chr.16: 21971652-21971652 on Build GRCh38
rs367686604 Chr.16: 21971536-21971536 on Build GRCh38
rs754265394 Chr.16: 21971569-21971569 on Build GRCh38
rs756357491 Chr.16: 21971537-21971537 on Build GRCh38
rs967790765 Chr.16: 21971524-21971524 on Build GRCh38
rs1023540852 Chr.16: 21971617-21971617 on Build GRCh38
rs201061280 Chr.16: 21971515-21971515 on Build GRCh38
rs372070369 Chr.16: 21971635-21971635 on Build GRCh38
rs867029147 Chr.16: 21971610-21971610 on Build GRCh38
rs778594446 Chr.16: 21971549-21971557 on Build GRCh38
rs192560484 Chr.16: 21971655-21971655 on Build GRCh38
rs772053496 Chr.16: 21971637-21971637 on Build GRCh38
rs573485610 Chr.16: 21971661-21971661 on Build GRCh38
rs772967686 Chr.16: 21971656-21971656 on Build GRCh38
rs766334618 Chr.16: 21971662-21971662 on Build GRCh38
rs905859276 Chr.16: 21971670-21971670 on Build GRCh38
rs780465182 Chr.16: 21971612-21971612 on Build GRCh38
rs779473664 Chr.16: 21971605-21971605 on Build GRCh38
rs371542784 Chr.16: 21971563-21971563 on Build GRCh38
rs769886992 Chr.16: 21971538-21971538 on Build GRCh38
rs779591300 Chr.16: 21971572-21971572 on Build GRCh38
rs757625414 Chr.16: 21971570-21971570 on Build GRCh38
rs767542991 Chr.16: 21971533-21971533 on Build GRCh38
rs1048190756 Chr.16: 21971513-21971513 on Build GRCh38
rs758919651 Chr.16: 21971577-21971577 on Build GRCh38
rs746331947 Chr.16: 21971574-21971574 on Build GRCh38
rs746526383 Chr.16: 21971614-21971614 on Build GRCh38
rs764361077 Chr.16: 21971542-21971542 on Build GRCh38

View Full Product Details