Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119358733-119359002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP2
Entrez Gene ID
9099
Gene Name
ubiquitin specific peptidase 2
Gene Aliases
UBP41, USP9
UniGene
Hs.524085
Gene Chromosome Location
Chr.11: 119355215-119381726 on Build GRCh38
Transcripts
NM_001243759.1 NM_004205.4 NM_171997.2 XM_005271721.4 XM_005271722.2 XM_017018539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809478 c.575G>C p.R192T Chr.11: 119358808-119358808 on Build GRCh38
1585444 c.606C>T p.I202I Chr.11: 119358777-119358777 on Build GRCh38
2106933 c.1222G>C p.D408H Chr.11: 119358788-119358788 on Build GRCh38
6498850 c.1216C>T p.R406W Chr.11: 119358794-119358794 on Build GRCh38
6818323 c.575G>T p.R192I Chr.11: 119358808-119358808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147093004 Chr.11: 119358804-119358804 on Build GRCh38
rs770081903 Chr.11: 119358855-119358855 on Build GRCh38
rs779603066 Chr.11: 119358836-119358836 on Build GRCh38
rs1020544967 Chr.11: 119358935-119358935 on Build GRCh38
rs781558102 Chr.11: 119358848-119358848 on Build GRCh38
rs201636268 Chr.11: 119358876-119358876 on Build GRCh38
rs140569658 Chr.11: 119358776-119358776 on Build GRCh38
rs45570535 Chr.11: 119358886-119358886 on Build GRCh38
rs764137919 Chr.11: 119358797-119358797 on Build GRCh38
rs374271839 Chr.11: 119358844-119358844 on Build GRCh38
rs761543830 Chr.11: 119358813-119358813 on Build GRCh38
rs943043880 Chr.11: 119358933-119358933 on Build GRCh38
rs904088505 Chr.11: 119358964-119358964 on Build GRCh38
rs888922783 Chr.11: 119358770-119358770 on Build GRCh38
rs369164717 Chr.11: 119358794-119358794 on Build GRCh38
rs200005535 Chr.11: 119358863-119358863 on Build GRCh38
rs749562448 Chr.11: 119358862-119358862 on Build GRCh38
rs764976766 Chr.11: 119358977-119358977 on Build GRCh38
rs371751796 Chr.11: 119358856-119358856 on Build GRCh38
rs553334685 Chr.11: 119358878-119358882 on Build GRCh38
rs775538652 Chr.11: 119358793-119358793 on Build GRCh38
rs752045077 Chr.11: 119358837-119358837 on Build GRCh38
rs768101363 Chr.11: 119358976-119358976 on Build GRCh38
rs759691426 Chr.11: 119358872-119358872 on Build GRCh38
rs778034274 Chr.11: 119358861-119358861 on Build GRCh38
rs778789064 Chr.11: 119358847-119358850 on Build GRCh38
rs145903800 Chr.11: 119358755-119358755 on Build GRCh38
rs970455800 Chr.11: 119358953-119358953 on Build GRCh38
rs764894922 Chr.11: 119358820-119358820 on Build GRCh38
rs771185028 Chr.11: 119358777-119358777 on Build GRCh38
rs774632637 Chr.11: 119358865-119358865 on Build GRCh38
rs748509129 Chr.11: 119358854-119358854 on Build GRCh38
rs757870112 Chr.11: 119358831-119358831 on Build GRCh38
rs773341497 Chr.11: 119358769-119358769 on Build GRCh38
rs769995020 Chr.11: 119358766-119358766 on Build GRCh38
rs1032700398 Chr.11: 119358869-119358869 on Build GRCh38
rs750087880 Chr.11: 119358830-119358830 on Build GRCh38

View Full Product Details