Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186946566-186946827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G4A
Entrez Gene ID
5321
Gene Name
phospholipase A2 group IVA
Gene Aliases
PLA2G4, cPLA2, cPLA2-alpha
UniGene
Hs.497200
Gene Chromosome Location
Chr.1: 186828900-186989896 on Build GRCh38
Transcripts
NM_001311193.1 NM_024420.2 XM_005245267.3 XM_011509642.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3734174 c.1059A>T p.E353D Chr.1: 186946662-186946662 on Build GRCh38
6390594 c.1073A>T p.K358I Chr.1: 186946676-186946676 on Build GRCh38
4458626 c.1093C>T p.P365S Chr.1: 186946696-186946696 on Build GRCh38
5697677 c.1095C>T p.P365P Chr.1: 186946698-186946698 on Build GRCh38
6009682 c.1078G>A p.G360S Chr.1: 186946681-186946681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200362578 Chr.1: 186946798-186946798 on Build GRCh38
rs760914662 Chr.1: 186946672-186946672 on Build GRCh38
rs1029275407 Chr.1: 186946694-186946694 on Build GRCh38
rs1018916737 Chr.1: 186946743-186946743 on Build GRCh38
rs781515658 Chr.1: 186946727-186946727 on Build GRCh38
rs757468267 Chr.1: 186946705-186946705 on Build GRCh38
rs763504723 Chr.1: 186946698-186946698 on Build GRCh38
rs970940557 Chr.1: 186946717-186946717 on Build GRCh38
rs753713596 Chr.1: 186946729-186946729 on Build GRCh38
rs747664337 Chr.1: 186946602-186946602 on Build GRCh38
rs572102047 Chr.1: 186946647-186946647 on Build GRCh38
rs764385879 Chr.1: 186946680-186946680 on Build GRCh38
rs868271147 Chr.1: 186946671-186946671 on Build GRCh38
rs772018373 Chr.1: 186946735-186946735 on Build GRCh38
rs776200451 Chr.1: 186946762-186946762 on Build GRCh38
rs779005617 Chr.1: 186946732-186946732 on Build GRCh38
rs770204210 Chr.1: 186946782-186946782 on Build GRCh38
rs139965148 Chr.1: 186946746-186946746 on Build GRCh38
rs546228398 Chr.1: 186946734-186946734 on Build GRCh38
rs374085425 Chr.1: 186946686-186946686 on Build GRCh38
rs750546323 Chr.1: 186946716-186946717 on Build GRCh38
rs77939449 Chr.1: 186946708-186946708 on Build GRCh38
rs752982663 Chr.1: 186946607-186946607 on Build GRCh38
rs770521684 Chr.1: 186946754-186946754 on Build GRCh38
rs759854632 Chr.1: 186946659-186946659 on Build GRCh38
rs746246855 Chr.1: 186946779-186946779 on Build GRCh38
rs770599762 Chr.1: 186946638-186946638 on Build GRCh38
rs777006571 Chr.1: 186946656-186946656 on Build GRCh38
rs369156087 Chr.1: 186946613-186946613 on Build GRCh38
rs751625866 Chr.1: 186946699-186946699 on Build GRCh38
rs760287431 Chr.1: 186946619-186946619 on Build GRCh38
rs754170276 Chr.1: 186946693-186946693 on Build GRCh38
rs778950514 Chr.1: 186946588-186946588 on Build GRCh38
rs754749824 Chr.1: 186946731-186946731 on Build GRCh38
rs1032822709 Chr.1: 186946636-186946636 on Build GRCh38
rs777201359 Chr.1: 186946740-186946740 on Build GRCh38
rs771422919 Chr.1: 186946605-186946605 on Build GRCh38

View Full Product Details