Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 54518475-54518714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX18
Entrez Gene ID
112574
Gene Name
sorting nexin 18
Gene Aliases
SH3PX2, SH3PXD3B, SNAG1
UniGene
Hs.432755
Gene Chromosome Location
Chr.5: 54517759-54619249 on Build GRCh38
Transcripts
NM_001102575.1 NM_001145427.1 NM_052870.2 XM_017008997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5621879 c.698C>T p.S233F Chr.5: 54518650-54518650 on Build GRCh38
3855565 c.706G>A p.V236I Chr.5: 54518658-54518658 on Build GRCh38
1068735 c.753C>T p.F251F Chr.5: 54518705-54518705 on Build GRCh38
6786440 c.754G>A p.V252M Chr.5: 54518706-54518706 on Build GRCh38
6103973 c.743C>T p.A248V Chr.5: 54518695-54518695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964311384 Chr.5: 54518548-54518548 on Build GRCh38
rs756900849 Chr.5: 54518606-54518606 on Build GRCh38
rs200257949 Chr.5: 54518522-54518522 on Build GRCh38
rs768296088 Chr.5: 54518681-54518681 on Build GRCh38
rs772468818 Chr.5: 54518497-54518497 on Build GRCh38
rs376563449 Chr.5: 54518648-54518648 on Build GRCh38
rs765919643 Chr.5: 54518563-54518564 on Build GRCh38
rs751054312 Chr.5: 54518564-54518565 on Build GRCh38
rs750232266 Chr.5: 54518544-54518544 on Build GRCh38
rs930396306 Chr.5: 54518579-54518579 on Build GRCh38
rs778312169 Chr.5: 54518659-54518659 on Build GRCh38
rs138425349 Chr.5: 54518690-54518690 on Build GRCh38
rs747434963 Chr.5: 54518660-54518660 on Build GRCh38
rs758456301 Chr.5: 54518654-54518654 on Build GRCh38
rs755181884 Chr.5: 54518645-54518645 on Build GRCh38
rs756679054 Chr.5: 54518649-54518649 on Build GRCh38
rs777012786 Chr.5: 54518587-54518587 on Build GRCh38
rs374567519 Chr.5: 54518542-54518542 on Build GRCh38
rs1052473199 Chr.5: 54518526-54518526 on Build GRCh38
rs760469131 Chr.5: 54518695-54518695 on Build GRCh38
rs531066338 Chr.5: 54518585-54518585 on Build GRCh38
rs772796832 Chr.5: 54518694-54518694 on Build GRCh38
rs780288239 Chr.5: 54518550-54518550 on Build GRCh38
rs771439275 Chr.5: 54518692-54518692 on Build GRCh38
rs779075343 Chr.5: 54518557-54518557 on Build GRCh38
rs746238651 Chr.5: 54518677-54518677 on Build GRCh38
rs935529153 Chr.5: 54518516-54518516 on Build GRCh38
rs773786121 Chr.5: 54518600-54518600 on Build GRCh38
rs201253342 Chr.5: 54518598-54518598 on Build GRCh38
rs751093332 Chr.5: 54518539-54518539 on Build GRCh38
rs199620482 Chr.5: 54518622-54518622 on Build GRCh38
rs748613247 Chr.5: 54518492-54518492 on Build GRCh38
rs755269988 Chr.5: 54518555-54518555 on Build GRCh38
rs141661757 Chr.5: 54518691-54518691 on Build GRCh38
rs1043889415 Chr.5: 54518495-54518495 on Build GRCh38
rs560095073 Chr.5: 54518507-54518507 on Build GRCh38
rs752809825 Chr.5: 54518651-54518651 on Build GRCh38
rs775170949 Chr.5: 54518510-54518510 on Build GRCh38
rs773556595 Chr.5: 54518506-54518506 on Build GRCh38
rs757806316 Chr.5: 54518666-54518666 on Build GRCh38
rs987085285 Chr.5: 54518525-54518525 on Build GRCh38
rs753695635 Chr.5: 54518627-54518627 on Build GRCh38
rs766223698 Chr.5: 54518624-54518624 on Build GRCh38
rs542228619 Chr.5: 54518545-54518545 on Build GRCh38
rs761300441 Chr.5: 54518607-54518607 on Build GRCh38
rs769472742 Chr.5: 54518508-54518508 on Build GRCh38
rs1050920542 Chr.5: 54518570-54518570 on Build GRCh38
rs778201349 Chr.5: 54518582-54518582 on Build GRCh38
rs371334283 Chr.5: 54518580-54518580 on Build GRCh38
rs761682032 Chr.5: 54518540-54518540 on Build GRCh38
rs2548614 Chr.5: 54518564-54518564 on Build GRCh38
rs368141304 Chr.5: 54518615-54518615 on Build GRCh38
rs920057148 Chr.5: 54518568-54518568 on Build GRCh38
rs905814644 Chr.5: 54518631-54518631 on Build GRCh38
rs754069192 Chr.5: 54518553-54518553 on Build GRCh38
rs762545075 Chr.5: 54518588-54518588 on Build GRCh38

View Full Product Details