Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20429641-20429866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARF2
Entrez Gene ID
91179
Gene Name
scavenger receptor class F member 2
Gene Aliases
NSR1, SREC-II, SREC2, SRECRP-1, VDEGS
UniGene
Hs.474251
Gene Chromosome Location
Chr.22: 20424584-20437859 on Build GRCh38
Transcripts
NM_153334.6 NM_182895.4 XM_017029065.1

SNP Detail

SNP ID Chromosome Location
rs760565659 Chr.22: 20429693-20429693 on Build GRCh38
rs759292519 Chr.22: 20429838-20429838 on Build GRCh38
rs775850255 Chr.22: 20429667-20429667 on Build GRCh38
rs765550586 Chr.22: 20429829-20429830 on Build GRCh38
rs752452819 Chr.22: 20429845-20429845 on Build GRCh38
rs372838078 Chr.22: 20429704-20429704 on Build GRCh38
rs952087770 Chr.22: 20429808-20429808 on Build GRCh38
rs1026866059 Chr.22: 20429803-20429803 on Build GRCh38
rs752432579 Chr.22: 20429707-20429707 on Build GRCh38
rs770130348 Chr.22: 20429666-20429666 on Build GRCh38
rs754824225 Chr.22: 20429697-20429697 on Build GRCh38
rs775910848 Chr.22: 20429837-20429838 on Build GRCh38
rs764283537 Chr.22: 20429683-20429683 on Build GRCh38
rs770235904 Chr.22: 20429770-20429770 on Build GRCh38
rs768792468 Chr.22: 20429800-20429800 on Build GRCh38
rs774347409 Chr.22: 20429812-20429812 on Build GRCh38
rs753570676 Chr.22: 20429696-20429696 on Build GRCh38
rs963886198 Chr.22: 20429730-20429730 on Build GRCh38
rs556727567 Chr.22: 20429720-20429720 on Build GRCh38
rs202155444 Chr.22: 20429699-20429699 on Build GRCh38
rs187721146 Chr.22: 20429673-20429673 on Build GRCh38
rs776615693 Chr.22: 20429822-20429822 on Build GRCh38
rs759996535 Chr.22: 20429663-20429663 on Build GRCh38
rs766288090 Chr.22: 20429818-20429818 on Build GRCh38
rs757785046 Chr.22: 20429761-20429761 on Build GRCh38
rs777073246 Chr.22: 20429661-20429661 on Build GRCh38
rs759982094 Chr.22: 20429707-20429708 on Build GRCh38
rs775554486 Chr.22: 20429776-20429776 on Build GRCh38
rs897620875 Chr.22: 20429772-20429772 on Build GRCh38
rs35216297 Chr.22: 20429815-20429815 on Build GRCh38
rs2241230 Chr.22: 20429763-20429763 on Build GRCh38
rs758025176 Chr.22: 20429849-20429849 on Build GRCh38
rs991426224 Chr.22: 20429662-20429662 on Build GRCh38
rs746233729 Chr.22: 20429766-20429766 on Build GRCh38
rs777284397 Chr.22: 20429734-20429734 on Build GRCh38
rs765126512 Chr.22: 20429839-20429839 on Build GRCh38
rs751614180 Chr.22: 20429691-20429691 on Build GRCh38
rs746526068 Chr.22: 20429752-20429752 on Build GRCh38
rs766308409 Chr.22: 20429694-20429694 on Build GRCh38
rs1049591052 Chr.22: 20429826-20429826 on Build GRCh38
rs749477721 Chr.22: 20429796-20429796 on Build GRCh38

View Full Product Details