Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56159222-56159473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8K5
Entrez Gene ID
219453
Gene Name
olfactory receptor family 8 subfamily K member 5
Gene Aliases
OR11-174
UniGene
Hs.553626
Gene Chromosome Location
Chr.11: 56159394-56160317 on Build GRCh38
Transcripts
NM_001004058.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
688658 c.886G>A p.E296K Chr.11: 56159432-56159432 on Build GRCh38
4510779 c.848C>T p.P283L Chr.11: 56159470-56159470 on Build GRCh38
6132965 c.877A>G p.R293G Chr.11: 56159441-56159441 on Build GRCh38
5743463 c.857A>T p.N286I Chr.11: 56159461-56159461 on Build GRCh38
7225657 c.903C>A p.F301L Chr.11: 56159415-56159415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020529127 Chr.11: 56159281-56159281 on Build GRCh38
rs141718394 Chr.11: 56159408-56159408 on Build GRCh38
rs1003188515 Chr.11: 56159249-56159249 on Build GRCh38
rs79877849 Chr.11: 56159364-56159364 on Build GRCh38
rs377062462 Chr.11: 56159383-56159383 on Build GRCh38
rs537183504 Chr.11: 56159335-56159335 on Build GRCh38
rs756503799 Chr.11: 56159371-56159371 on Build GRCh38
rs761707199 Chr.11: 56159350-56159350 on Build GRCh38
rs775649734 Chr.11: 56159412-56159412 on Build GRCh38
rs1048792978 Chr.11: 56159446-56159446 on Build GRCh38
rs559349522 Chr.11: 56159289-56159289 on Build GRCh38
rs530593244 Chr.11: 56159369-56159369 on Build GRCh38
rs2512774 Chr.11: 56159320-56159320 on Build GRCh38
rs768418948 Chr.11: 56159419-56159419 on Build GRCh38
rs943857277 Chr.11: 56159388-56159388 on Build GRCh38
rs577105438 Chr.11: 56159256-56159256 on Build GRCh38
rs766014059 Chr.11: 56159445-56159445 on Build GRCh38
rs777587934 Chr.11: 56159310-56159310 on Build GRCh38
rs931272029 Chr.11: 56159444-56159444 on Build GRCh38
rs979163044 Chr.11: 56159302-56159302 on Build GRCh38
rs138593758 Chr.11: 56159406-56159406 on Build GRCh38
rs772152342 Chr.11: 56159411-56159411 on Build GRCh38
rs760911007 Chr.11: 56159413-56159413 on Build GRCh38
rs761807276 Chr.11: 56159434-56159434 on Build GRCh38
rs771380091 Chr.11: 56159395-56159395 on Build GRCh38
rs540960525 Chr.11: 56159282-56159282 on Build GRCh38
rs75866237 Chr.11: 56159329-56159329 on Build GRCh38
rs751258224 Chr.11: 56159370-56159370 on Build GRCh38
rs1010426782 Chr.11: 56159275-56159275 on Build GRCh38
rs751205072 Chr.11: 56159448-56159448 on Build GRCh38
rs1011542100 Chr.11: 56159311-56159311 on Build GRCh38
rs749971676 Chr.11: 56159359-56159359 on Build GRCh38
rs772325896 Chr.11: 56159435-56159435 on Build GRCh38
rs558442461 Chr.11: 56159250-56159250 on Build GRCh38
rs376114385 Chr.11: 56159435-56159435 on Build GRCh38
rs921892584 Chr.11: 56159373-56159373 on Build GRCh38
rs764776732 Chr.11: 56159354-56159354 on Build GRCh38
rs147115733 Chr.11: 56159436-56159436 on Build GRCh38
rs776333511 Chr.11: 56159426-56159426 on Build GRCh38

View Full Product Details