Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57301588-57301853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNKS1BP1
Entrez Gene ID
85456
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
TAB182
UniGene
Hs.530730
Gene Chromosome Location
Chr.11: 57299629-57324952 on Build GRCh38
Transcripts
NM_033396.2 XM_006718725.3

SNP Detail

SNP ID Chromosome Location
rs1053885393 Chr.11: 57301678-57301678 on Build GRCh38
rs905174389 Chr.11: 57301640-57301640 on Build GRCh38
rs184100518 Chr.11: 57301799-57301799 on Build GRCh38
rs565483000 Chr.11: 57301747-57301747 on Build GRCh38
rs759381339 Chr.11: 57301825-57301833 on Build GRCh38
rs950339176 Chr.11: 57301814-57301814 on Build GRCh38
rs141088269 Chr.11: 57301683-57301683 on Build GRCh38
rs555132411 Chr.11: 57301660-57301660 on Build GRCh38
rs772877226 Chr.11: 57301765-57301765 on Build GRCh38
rs780473051 Chr.11: 57301828-57301828 on Build GRCh38
rs1008756208 Chr.11: 57301739-57301739 on Build GRCh38
rs767298654 Chr.11: 57301793-57301793 on Build GRCh38
rs750744427 Chr.11: 57301813-57301813 on Build GRCh38
rs890403187 Chr.11: 57301736-57301736 on Build GRCh38
rs991882344 Chr.11: 57301697-57301697 on Build GRCh38
rs752446811 Chr.11: 57301796-57301796 on Build GRCh38
rs543083757 Chr.11: 57301625-57301625 on Build GRCh38
rs770933673 Chr.11: 57301773-57301773 on Build GRCh38
rs369491793 Chr.11: 57301772-57301772 on Build GRCh38
rs202093593 Chr.11: 57301785-57301785 on Build GRCh38
rs759073193 Chr.11: 57301792-57301792 on Build GRCh38
rs1017249598 Chr.11: 57301744-57301744 on Build GRCh38
rs757211340 Chr.11: 57301620-57301620 on Build GRCh38
rs776393935 Chr.11: 57301759-57301761 on Build GRCh38
rs1046460861 Chr.11: 57301638-57301638 on Build GRCh38
rs909943623 Chr.11: 57301669-57301669 on Build GRCh38
rs868119122 Chr.11: 57301641-57301641 on Build GRCh38
rs758920354 Chr.11: 57301815-57301815 on Build GRCh38
rs949652154 Chr.11: 57301634-57301635 on Build GRCh38
rs765703525 Chr.11: 57301812-57301812 on Build GRCh38
rs763041855 Chr.11: 57301766-57301766 on Build GRCh38

View Full Product Details