Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58498904-58499177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A5
Entrez Gene ID
10998
Gene Name
solute carrier family 27 member 5
Gene Aliases
ACSB, ACSVL6, BACS, BAL, FACVL3, FATP-5, FATP5, VLACSR, VLCS-H2, VLCSH2
UniGene
Hs.292177
Gene Chromosome Location
Chr.19: 58498333-58512065 on Build GRCh38
Transcripts
NM_001321196.1 NM_012254.2 XM_011526364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854730 c.1769G>C p.C590S Chr.19: 58498912-58498912 on Build GRCh38
4412942 c.1743C>G p.N581K Chr.19: 58499145-58499145 on Build GRCh38
6152495 c.1763C>A p.P588Q Chr.19: 58499125-58499125 on Build GRCh38
4427643 c.1754T>A p.V585E Chr.19: 58499134-58499134 on Build GRCh38
4975348 c.1774G>A p.G592S Chr.19: 58498907-58498907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905173513 Chr.19: 58499057-58499057 on Build GRCh38
rs761352933 Chr.19: 58499078-58499078 on Build GRCh38
rs375604806 Chr.19: 58499091-58499091 on Build GRCh38
rs752816081 Chr.19: 58499093-58499093 on Build GRCh38
rs757262114 Chr.19: 58498933-58498934 on Build GRCh38
rs80330258 Chr.19: 58499072-58499072 on Build GRCh38
rs771502444 Chr.19: 58499136-58499136 on Build GRCh38
rs1003518200 Chr.19: 58499062-58499062 on Build GRCh38
rs754023114 Chr.19: 58498932-58498932 on Build GRCh38
rs747812229 Chr.19: 58499126-58499126 on Build GRCh38
rs745648383 Chr.19: 58498965-58498965 on Build GRCh38
rs745430167 Chr.19: 58499147-58499148 on Build GRCh38
rs201952004 Chr.19: 58499074-58499074 on Build GRCh38
rs748932500 Chr.19: 58499073-58499073 on Build GRCh38
rs780829121 Chr.19: 58498941-58498964 on Build GRCh38
rs372050476 Chr.19: 58499120-58499120 on Build GRCh38
rs757226749 Chr.19: 58498959-58498959 on Build GRCh38
rs765593428 Chr.19: 58499092-58499092 on Build GRCh38
rs773830618 Chr.19: 58499076-58499076 on Build GRCh38
rs111249515 Chr.19: 58499032-58499032 on Build GRCh38
rs566996803 Chr.19: 58498995-58498995 on Build GRCh38
rs933659666 Chr.19: 58498978-58498978 on Build GRCh38
rs370943108 Chr.19: 58499022-58499022 on Build GRCh38
rs772739623 Chr.19: 58499139-58499139 on Build GRCh38
rs755185254 Chr.19: 58498939-58498939 on Build GRCh38
rs368083874 Chr.19: 58499114-58499114 on Build GRCh38
rs190235885 Chr.19: 58499090-58499090 on Build GRCh38
rs899366632 Chr.19: 58499043-58499043 on Build GRCh38
rs777020403 Chr.19: 58499154-58499154 on Build GRCh38
rs912152408 Chr.19: 58499013-58499013 on Build GRCh38
rs559823637 Chr.19: 58498982-58498982 on Build GRCh38
rs375962515 Chr.19: 58499140-58499140 on Build GRCh38
rs373860145 Chr.19: 58498937-58498937 on Build GRCh38
rs745736118 Chr.19: 58499103-58499103 on Build GRCh38
rs761090069 Chr.19: 58499084-58499084 on Build GRCh38
rs77807864 Chr.19: 58499157-58499157 on Build GRCh38
rs994910692 Chr.19: 58499046-58499046 on Build GRCh38
rs773650953 Chr.19: 58499083-58499083 on Build GRCh38
rs528553647 Chr.19: 58499003-58499003 on Build GRCh38
rs748284017 Chr.19: 58498958-58498958 on Build GRCh38
rs771388335 Chr.19: 58499145-58499145 on Build GRCh38
rs758614063 Chr.19: 58499097-58499097 on Build GRCh38
rs756038579 Chr.19: 58499113-58499113 on Build GRCh38
rs200170382 Chr.19: 58498926-58498926 on Build GRCh38
rs368570614 Chr.19: 58499082-58499082 on Build GRCh38
rs778016405 Chr.19: 58499102-58499102 on Build GRCh38
rs1006837339 Chr.19: 58498954-58498954 on Build GRCh38
rs778999156 Chr.19: 58498956-58498956 on Build GRCh38
rs999768492 Chr.19: 58499035-58499035 on Build GRCh38
rs940821708 Chr.19: 58499018-58499018 on Build GRCh38
rs571438946 Chr.19: 58499052-58499052 on Build GRCh38
rs774057315 Chr.19: 58499124-58499124 on Build GRCh38

View Full Product Details