Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13708715-13708970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB9A
Entrez Gene ID
9367
Gene Name
RAB9A, member RAS oncogene family
Gene Aliases
RAB9
UniGene
Hs.495704
Gene Chromosome Location
Chr.X: 13689121-13709825 on Build GRCh38
Transcripts
NM_001195328.1 NM_004251.4 XM_017029964.1

SNP Detail

SNP ID Chromosome Location
rs747605998 Chr.X: 13708935-13708935 on Build GRCh38
rs748175382 Chr.X: 13708941-13708941 on Build GRCh38
rs753858871 Chr.X: 13708788-13708788 on Build GRCh38
rs878947803 Chr.X: 13708866-13708866 on Build GRCh38
rs757540588 Chr.X: 13708839-13708839 on Build GRCh38
rs780502129 Chr.X: 13708907-13708907 on Build GRCh38
rs779259114 Chr.X: 13708900-13708900 on Build GRCh38
rs776240786 Chr.X: 13708943-13708943 on Build GRCh38
rs56269332 Chr.X: 13708821-13708821 on Build GRCh38
rs776898712 Chr.X: 13708867-13708867 on Build GRCh38
rs1035127001 Chr.X: 13708893-13708893 on Build GRCh38
rs772597973 Chr.X: 13708906-13708906 on Build GRCh38
rs754816289 Chr.X: 13708759-13708759 on Build GRCh38
rs11542996 Chr.X: 13708761-13708761 on Build GRCh38
rs751501956 Chr.X: 13708755-13708755 on Build GRCh38
rs1023049780 Chr.X: 13708908-13708908 on Build GRCh38
rs746042668 Chr.X: 13708905-13708905 on Build GRCh38

View Full Product Details