Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27810129-27810402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX12
Entrez Gene ID
23673
Gene Name
syntaxin 12
Gene Aliases
STX13, STX14
UniGene
Hs.523855
Gene Chromosome Location
Chr.1: 27773183-27824452 on Build GRCh38
Transcripts
NM_177424.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4730718 c.470+2T>A p.? Chr.1: 27810291-27810291 on Build GRCh38
4730719 c.470+2T>A p.? Chr.1: 27810291-27810291 on Build GRCh38
4751740 c.470G>A p.S157N Chr.1: 27810289-27810289 on Build GRCh38
3771711 c.467A>T p.D156V Chr.1: 27810286-27810286 on Build GRCh38
3771712 c.467A>T p.D156V Chr.1: 27810286-27810286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531550444 Chr.1: 27810308-27810308 on Build GRCh38
rs1007612319 Chr.1: 27810257-27810257 on Build GRCh38
rs773716755 Chr.1: 27810317-27810317 on Build GRCh38
rs752791985 Chr.1: 27810282-27810282 on Build GRCh38
rs745482883 Chr.1: 27810271-27810271 on Build GRCh38
rs79262050 Chr.1: 27810374-27810374 on Build GRCh38
rs775695594 Chr.1: 27810285-27810285 on Build GRCh38
rs766643924 Chr.1: 27810173-27810173 on Build GRCh38
rs769173899 Chr.1: 27810294-27810294 on Build GRCh38
rs779559361 Chr.1: 27810283-27810283 on Build GRCh38
rs1036599660 Chr.1: 27810159-27810159 on Build GRCh38
rs1027792781 Chr.1: 27810181-27810181 on Build GRCh38
rs756385921 Chr.1: 27810309-27810309 on Build GRCh38
rs951549041 Chr.1: 27810370-27810370 on Build GRCh38
rs778705939 Chr.1: 27810238-27810238 on Build GRCh38
rs186053587 Chr.1: 27810381-27810381 on Build GRCh38
rs376491372 Chr.1: 27810226-27810226 on Build GRCh38
rs955479272 Chr.1: 27810272-27810272 on Build GRCh38
rs543446421 Chr.1: 27810362-27810362 on Build GRCh38
rs766566932 Chr.1: 27810339-27810339 on Build GRCh38
rs17162805 Chr.1: 27810296-27810296 on Build GRCh38
rs999393739 Chr.1: 27810192-27810192 on Build GRCh38
rs977431216 Chr.1: 27810293-27810293 on Build GRCh38
rs1019473581 Chr.1: 27810266-27810266 on Build GRCh38
rs548198465 Chr.1: 27810330-27810330 on Build GRCh38
rs953689787 Chr.1: 27810206-27810206 on Build GRCh38
rs771629228 Chr.1: 27810280-27810280 on Build GRCh38
rs532643204 Chr.1: 27810156-27810156 on Build GRCh38
rs376238269 Chr.1: 27810343-27810343 on Build GRCh38
rs770240362 Chr.1: 27810299-27810299 on Build GRCh38
rs753601582 Chr.1: 27810218-27810218 on Build GRCh38
rs898121728 Chr.1: 27810163-27810163 on Build GRCh38
rs746560332 Chr.1: 27810289-27810289 on Build GRCh38
rs368249119 Chr.1: 27810260-27810260 on Build GRCh38

View Full Product Details