Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141364653-141364912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_032054.1 NM_018922.2 NM_018923.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5702354 c.479_480CG>AC p.A160D Chr.5: 141364809-141364810 on Build GRCh38
5167853 c.400C>T p.R134W Chr.5: 141364730-141364730 on Build GRCh38
6347885 c.469G>T p.A157S Chr.5: 141364799-141364799 on Build GRCh38
3851803 c.503G>A p.R168Q Chr.5: 141364833-141364833 on Build GRCh38
6351935 c.498T>C p.S166S Chr.5: 141364828-141364828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746438679 Chr.5: 141364723-141364723 on Build GRCh38
rs779536020 Chr.5: 141364722-141364722 on Build GRCh38
rs772314048 Chr.5: 141364724-141364724 on Build GRCh38
rs755884627 Chr.5: 141364769-141364769 on Build GRCh38
rs779195716 Chr.5: 141364809-141364809 on Build GRCh38
rs377438861 Chr.5: 141364705-141364705 on Build GRCh38
rs57308563 Chr.5: 141364828-141364828 on Build GRCh38
rs774257321 Chr.5: 141364741-141364741 on Build GRCh38
rs200583878 Chr.5: 141364836-141364836 on Build GRCh38
rs775321480 Chr.5: 141364871-141364871 on Build GRCh38
rs199575653 Chr.5: 141364858-141364858 on Build GRCh38
rs762418103 Chr.5: 141364738-141364738 on Build GRCh38
rs765921819 Chr.5: 141364739-141364739 on Build GRCh38
rs536236780 Chr.5: 141364733-141364733 on Build GRCh38
rs909012191 Chr.5: 141364875-141364875 on Build GRCh38
rs772829595 Chr.5: 141364855-141364855 on Build GRCh38
rs757392187 Chr.5: 141364799-141364799 on Build GRCh38
rs748996283 Chr.5: 141364856-141364856 on Build GRCh38
rs142073719 Chr.5: 141364854-141364854 on Build GRCh38
rs1002059027 Chr.5: 141364873-141364873 on Build GRCh38
rs764374918 Chr.5: 141364772-141364772 on Build GRCh38
rs778155346 Chr.5: 141364710-141364710 on Build GRCh38
rs749361969 Chr.5: 141364866-141364866 on Build GRCh38
rs777714340 Chr.5: 141364816-141364816 on Build GRCh38
rs796800411 Chr.5: 141364790-141364790 on Build GRCh38
rs377017789 Chr.5: 141364766-141364766 on Build GRCh38
rs762196264 Chr.5: 141364884-141364884 on Build GRCh38
rs368317740 Chr.5: 141364874-141364874 on Build GRCh38
rs373731815 Chr.5: 141364691-141364691 on Build GRCh38
rs538468873 Chr.5: 141364832-141364832 on Build GRCh38
rs572371084 Chr.5: 141364847-141364847 on Build GRCh38
rs374170303 Chr.5: 141364730-141364730 on Build GRCh38
rs868142397 Chr.5: 141364767-141364767 on Build GRCh38
rs566972515 Chr.5: 141364715-141364715 on Build GRCh38
rs897458319 Chr.5: 141364818-141364818 on Build GRCh38
rs940260607 Chr.5: 141364754-141364754 on Build GRCh38
rs760519562 Chr.5: 141364872-141364872 on Build GRCh38
rs199576947 Chr.5: 141364882-141364882 on Build GRCh38
rs370237297 Chr.5: 141364711-141364711 on Build GRCh38
rs1036037278 Chr.5: 141364779-141364779 on Build GRCh38
rs754705935 Chr.5: 141364690-141364690 on Build GRCh38
rs775854228 Chr.5: 141364725-141364725 on Build GRCh38
rs558676002 Chr.5: 141364846-141364846 on Build GRCh38
rs767440295 Chr.5: 141364870-141364870 on Build GRCh38
rs891416528 Chr.5: 141364748-141364748 on Build GRCh38
rs932792949 Chr.5: 141364731-141364731 on Build GRCh38
rs549647263 Chr.5: 141364755-141364755 on Build GRCh38
rs765566914 Chr.5: 141364890-141364890 on Build GRCh38
rs754157065 Chr.5: 141364781-141364781 on Build GRCh38
rs767363057 Chr.5: 141364765-141364765 on Build GRCh38

View Full Product Details