Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46758901-46759128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS5
Entrez Gene ID
9655
Gene Name
suppressor of cytokine signaling 5
Gene Aliases
CIS6, CISH6, Cish5, SOCS-5
UniGene
Hs.468426
Gene Chromosome Location
Chr.2: 46698960-46762788 on Build GRCh38
Transcripts
NM_014011.4 NM_144949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3185736 c.448C>T p.Q150* Chr.2: 46758978-46758978 on Build GRCh38
6259995 c.427G>T p.G143C Chr.2: 46758957-46758957 on Build GRCh38
5730630 c.436_437insG p.S147fs*59 Chr.2: 46758966-46758967 on Build GRCh38
4833303 c.573G>C p.M191I Chr.2: 46759103-46759103 on Build GRCh38
4500660 c.570C>T p.P190P Chr.2: 46759100-46759100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756458628 Chr.2: 46759089-46759089 on Build GRCh38
rs764498188 Chr.2: 46759072-46759072 on Build GRCh38
rs776060821 Chr.2: 46759059-46759059 on Build GRCh38
rs757917355 Chr.2: 46759103-46759103 on Build GRCh38
rs770658440 Chr.2: 46759051-46759051 on Build GRCh38
rs754277862 Chr.2: 46759101-46759101 on Build GRCh38
rs781567392 Chr.2: 46759019-46759019 on Build GRCh38
rs544775730 Chr.2: 46759042-46759042 on Build GRCh38
rs752991722 Chr.2: 46758929-46758929 on Build GRCh38
rs759558091 Chr.2: 46758930-46758930 on Build GRCh38
rs780519623 Chr.2: 46759032-46759032 on Build GRCh38
rs759469437 Chr.2: 46758999-46758999 on Build GRCh38
rs770210992 Chr.2: 46759004-46759004 on Build GRCh38
rs544174086 Chr.2: 46759014-46759014 on Build GRCh38
rs140618503 Chr.2: 46759037-46759037 on Build GRCh38
rs771763816 Chr.2: 46758989-46758989 on Build GRCh38
rs781554212 Chr.2: 46758958-46758958 on Build GRCh38
rs138131726 Chr.2: 46759063-46759063 on Build GRCh38
rs760457146 Chr.2: 46758994-46758994 on Build GRCh38
rs764379487 Chr.2: 46759002-46759003 on Build GRCh38
rs778116720 Chr.2: 46758936-46758936 on Build GRCh38
rs757636569 Chr.2: 46758948-46758948 on Build GRCh38
rs779779913 Chr.2: 46759057-46759057 on Build GRCh38
rs750922089 Chr.2: 46759029-46759029 on Build GRCh38
rs767602215 Chr.2: 46759070-46759070 on Build GRCh38
rs577577930 Chr.2: 46759061-46759061 on Build GRCh38
rs758655262 Chr.2: 46758931-46758931 on Build GRCh38
rs189285831 Chr.2: 46759043-46759043 on Build GRCh38
rs143799189 Chr.2: 46758963-46758963 on Build GRCh38
rs774459211 Chr.2: 46759064-46759064 on Build GRCh38
rs770697781 Chr.2: 46758995-46758995 on Build GRCh38
rs148061519 Chr.2: 46759073-46759073 on Build GRCh38
rs766940328 Chr.2: 46759091-46759091 on Build GRCh38
rs762054405 Chr.2: 46759069-46759069 on Build GRCh38
rs576800210 Chr.2: 46758998-46758998 on Build GRCh38
rs267599392 Chr.2: 46758934-46758934 on Build GRCh38
rs987726251 Chr.2: 46759100-46759100 on Build GRCh38
rs769338868 Chr.2: 46759090-46759090 on Build GRCh38
rs772841848 Chr.2: 46758990-46758990 on Build GRCh38
rs751736443 Chr.2: 46759007-46759007 on Build GRCh38
rs373780921 Chr.2: 46759002-46759002 on Build GRCh38
rs752866954 Chr.2: 46759001-46759001 on Build GRCh38
rs756650688 Chr.2: 46759030-46759030 on Build GRCh38
rs751925296 Chr.2: 46758945-46758945 on Build GRCh38
rs202138086 Chr.2: 46759017-46759017 on Build GRCh38
rs768070642 Chr.2: 46758967-46758967 on Build GRCh38
rs757902963 Chr.2: 46759036-46759036 on Build GRCh38
rs747624486 Chr.2: 46758978-46758978 on Build GRCh38
rs745622402 Chr.2: 46759054-46759054 on Build GRCh38
rs776450161 Chr.2: 46759053-46759053 on Build GRCh38

View Full Product Details