Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120200665-120200910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPLP0
Entrez Gene ID
6175
Gene Name
ribosomal protein lateral stalk subunit P0
Gene Aliases
L10E, LP0, P0, PRLP0, RPP0
UniGene
Hs.546285
Gene Chromosome Location
Chr.12: 120196700-120201211 on Build GRCh38
Transcripts
NM_001002.3 NM_053275.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7195453 c.5C>T p.P2L Chr.12: 120200779-120200779 on Build GRCh38
5946727 c.27G>T p.W9C Chr.12: 120200757-120200757 on Build GRCh38
4494796 c.43C>T p.L15F Chr.12: 120200741-120200741 on Build GRCh38
3954283 c.2T>A p.M1K Chr.12: 120200782-120200782 on Build GRCh38
935889 c.25T>C p.W9R Chr.12: 120200759-120200759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111332516 Chr.12: 120200854-120200854 on Build GRCh38
rs888835947 Chr.12: 120200780-120200780 on Build GRCh38
rs756321565 Chr.12: 120200834-120200834 on Build GRCh38
rs1044953330 Chr.12: 120200846-120200846 on Build GRCh38
rs774101874 Chr.12: 120200737-120200737 on Build GRCh38
rs960177230 Chr.12: 120200837-120200837 on Build GRCh38
rs745835839 Chr.12: 120200742-120200742 on Build GRCh38
rs986455750 Chr.12: 120200767-120200767 on Build GRCh38
rs924590052 Chr.12: 120200797-120200797 on Build GRCh38
rs776275100 Chr.12: 120200794-120200794 on Build GRCh38
rs758958752 Chr.12: 120200695-120200695 on Build GRCh38
rs376911813 Chr.12: 120200815-120200815 on Build GRCh38
rs368912362 Chr.12: 120200791-120200791 on Build GRCh38
rs535872908 Chr.12: 120200723-120200723 on Build GRCh38
rs750876476 Chr.12: 120200789-120200789 on Build GRCh38
rs760219936 Chr.12: 120200763-120200763 on Build GRCh38
rs969167705 Chr.12: 120200847-120200847 on Build GRCh38
rs374029073 Chr.12: 120200800-120200800 on Build GRCh38
rs752111632 Chr.12: 120200707-120200707 on Build GRCh38
rs755547521 Chr.12: 120200818-120200818 on Build GRCh38
rs145990077 Chr.12: 120200849-120200849 on Build GRCh38
rs372714831 Chr.12: 120200730-120200730 on Build GRCh38
rs781644407 Chr.12: 120200713-120200713 on Build GRCh38
rs948873913 Chr.12: 120200829-120200829 on Build GRCh38
rs779931470 Chr.12: 120200730-120200732 on Build GRCh38
rs191903910 Chr.12: 120200715-120200715 on Build GRCh38
rs771721286 Chr.12: 120200746-120200746 on Build GRCh38
rs910509281 Chr.12: 120200778-120200778 on Build GRCh38
rs1013060554 Chr.12: 120200845-120200845 on Build GRCh38
rs370967854 Chr.12: 120200694-120200694 on Build GRCh38
rs764853143 Chr.12: 120200810-120200810 on Build GRCh38
rs763663208 Chr.12: 120200692-120200692 on Build GRCh38
rs780771891 Chr.12: 120200704-120200704 on Build GRCh38
rs748289315 Chr.12: 120200714-120200714 on Build GRCh38
rs747992508 Chr.12: 120200841-120200841 on Build GRCh38
rs184149210 Chr.12: 120200822-120200822 on Build GRCh38
rs142599353 Chr.12: 120200712-120200712 on Build GRCh38
rs1027920035 Chr.12: 120200733-120200733 on Build GRCh38
rs375856624 Chr.12: 120200749-120200749 on Build GRCh38
rs369200987 Chr.12: 120200719-120200719 on Build GRCh38
rs781520240 Chr.12: 120200855-120200855 on Build GRCh38
rs761341798 Chr.12: 120200809-120200809 on Build GRCh38
rs753160636 Chr.12: 120200825-120200825 on Build GRCh38

View Full Product Details