Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151760619-151760892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL9
Entrez Gene ID
65005
Gene Name
mitochondrial ribosomal protein L9
Gene Aliases
L9mt
UniGene
Hs.288936
Gene Chromosome Location
Chr.1: 151759643-151763916 on Build GRCh38
Transcripts
NM_001300733.1 NM_031420.3 NR_125331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022284 c.645G>A p.R215R Chr.1: 151760843-151760843 on Build GRCh38
3750355 c.637A>G p.I213V Chr.1: 151760851-151760851 on Build GRCh38
6715432 c.666G>A p.E222E Chr.1: 151760822-151760822 on Build GRCh38
895902 c.651C>T p.G217G Chr.1: 151760837-151760837 on Build GRCh38
5558924 c.617T>C p.L206S Chr.1: 151760871-151760871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020908618 Chr.1: 151760756-151760756 on Build GRCh38
rs370570047 Chr.1: 151760799-151760799 on Build GRCh38
rs780254266 Chr.1: 151760867-151760868 on Build GRCh38
rs773895639 Chr.1: 151760784-151760784 on Build GRCh38
rs900553726 Chr.1: 151760666-151760666 on Build GRCh38
rs74228558 Chr.1: 151760851-151760851 on Build GRCh38
rs377453943 Chr.1: 151760796-151760796 on Build GRCh38
rs750280308 Chr.1: 151760784-151760785 on Build GRCh38
rs551627980 Chr.1: 151760780-151760784 on Build GRCh38
rs539663039 Chr.1: 151760816-151760816 on Build GRCh38
rs753788923 Chr.1: 151760767-151760767 on Build GRCh38
rs144746208 Chr.1: 151760662-151760662 on Build GRCh38
rs780403884 Chr.1: 151760842-151760842 on Build GRCh38
rs142753752 Chr.1: 151760817-151760817 on Build GRCh38
rs779346768 Chr.1: 151760834-151760834 on Build GRCh38
rs979213255 Chr.1: 151760722-151760722 on Build GRCh38
rs568588737 Chr.1: 151760685-151760685 on Build GRCh38
rs139753069 Chr.1: 151760821-151760821 on Build GRCh38
rs774195497 Chr.1: 151760868-151760868 on Build GRCh38
rs557374486 Chr.1: 151760706-151760706 on Build GRCh38
rs79198441 Chr.1: 151760686-151760686 on Build GRCh38
rs1020837623 Chr.1: 151760687-151760687 on Build GRCh38
rs8480 Chr.1: 151760859-151760859 on Build GRCh38
rs374978268 Chr.1: 151760845-151760845 on Build GRCh38
rs761605014 Chr.1: 151760777-151760778 on Build GRCh38
rs755785195 Chr.1: 151760806-151760806 on Build GRCh38
rs989385373 Chr.1: 151760752-151760752 on Build GRCh38
rs572633637 Chr.1: 151760840-151760840 on Build GRCh38
rs769763380 Chr.1: 151760771-151760771 on Build GRCh38
rs766237242 Chr.1: 151760781-151760781 on Build GRCh38
rs758250657 Chr.1: 151760866-151760867 on Build GRCh38
rs752394385 Chr.1: 151760802-151760802 on Build GRCh38
rs773168837 Chr.1: 151760772-151760772 on Build GRCh38
rs755154290 Chr.1: 151760837-151760837 on Build GRCh38
rs139923281 Chr.1: 151760779-151760779 on Build GRCh38
rs545715070 Chr.1: 151760844-151760844 on Build GRCh38
rs777547766 Chr.1: 151760843-151760843 on Build GRCh38
rs957895153 Chr.1: 151760737-151760737 on Build GRCh38
rs949040930 Chr.1: 151760822-151760822 on Build GRCh38

View Full Product Details