Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231712669-231712938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTMA
Entrez Gene ID
5757
Gene Name
prothymosin, alpha
Gene Aliases
TMSA
UniGene
Hs.459927
Gene Chromosome Location
Chr.2: 231708525-231713541 on Build GRCh38
Transcripts
NM_001099285.1 NM_002823.4 XM_011511518.1 XM_011511519.1 XM_011511520.1 XM_011511521.1 XM_011511522.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6244916 c.298G>T p.D100Y Chr.2: 231712816-231712816 on Build GRCh38
1129284 c.291C>T p.D97D Chr.2: 231712809-231712809 on Build GRCh38
6238395 c.322G>A p.E108K Chr.2: 231712840-231712840 on Build GRCh38
1017775 c.298_311del14 p.D100fs*6 Chr.2: 231712816-231712829 on Build GRCh38
573838 c.331T>C p.*111Q Chr.2: 231712849-231712849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980241262 Chr.2: 231712810-231712810 on Build GRCh38
rs551638299 Chr.2: 231712842-231712842 on Build GRCh38
rs760419505 Chr.2: 231712868-231712868 on Build GRCh38
rs753400853 Chr.2: 231712862-231712863 on Build GRCh38
rs760052328 Chr.2: 231712791-231712791 on Build GRCh38
rs775557451 Chr.2: 231712836-231712836 on Build GRCh38
rs1804250 Chr.2: 231712809-231712809 on Build GRCh38
rs62197232 Chr.2: 231712889-231712889 on Build GRCh38
rs1031941273 Chr.2: 231712794-231712794 on Build GRCh38
rs750767589 Chr.2: 231712866-231712866 on Build GRCh38
rs753332066 Chr.2: 231712806-231712806 on Build GRCh38
rs755790774 Chr.2: 231712788-231712788 on Build GRCh38
rs767973983 Chr.2: 231712764-231712764 on Build GRCh38
rs200020726 Chr.2: 231712815-231712815 on Build GRCh38
rs563464274 Chr.2: 231712886-231712886 on Build GRCh38
rs921794203 Chr.2: 231712853-231712853 on Build GRCh38
rs573759409 Chr.2: 231712779-231712779 on Build GRCh38
rs758273484 Chr.2: 231712887-231712888 on Build GRCh38
rs903546344 Chr.2: 231712747-231712747 on Build GRCh38
rs777934444 Chr.2: 231712790-231712790 on Build GRCh38
rs779733063 Chr.2: 231712796-231712796 on Build GRCh38
rs879217836 Chr.2: 231712799-231712799 on Build GRCh38
rs1054738751 Chr.2: 231712869-231712869 on Build GRCh38
rs932215300 Chr.2: 231712862-231712863 on Build GRCh38
rs878990114 Chr.2: 231712801-231712801 on Build GRCh38
rs762198164 Chr.2: 231712849-231712849 on Build GRCh38
rs758287487 Chr.2: 231712892-231712892 on Build GRCh38
rs754572320 Chr.2: 231712882-231712882 on Build GRCh38
rs753427430 Chr.2: 231712877-231712877 on Build GRCh38
rs902347899 Chr.2: 231712719-231712719 on Build GRCh38
rs766063352 Chr.2: 231712876-231712876 on Build GRCh38
rs931789374 Chr.2: 231712891-231712891 on Build GRCh38
rs768608674 Chr.2: 231712839-231712839 on Build GRCh38
rs368475673 Chr.2: 231712763-231712763 on Build GRCh38
rs866602914 Chr.2: 231712687-231712687 on Build GRCh38
rs11558918 Chr.2: 231712837-231712837 on Build GRCh38
rs778264619 Chr.2: 231712884-231712884 on Build GRCh38
rs763706740 Chr.2: 231712815-231712816 on Build GRCh38
rs750184715 Chr.2: 231712880-231712881 on Build GRCh38
rs1014378920 Chr.2: 231712797-231712797 on Build GRCh38
rs13022221 Chr.2: 231712895-231712895 on Build GRCh38
rs202209669 Chr.2: 231712816-231712816 on Build GRCh38
rs1012194233 Chr.2: 231712778-231712778 on Build GRCh38
rs756999912 Chr.2: 231712877-231712877 on Build GRCh38
rs768026663 Chr.2: 231712864-231712864 on Build GRCh38
rs777564345 Chr.2: 231712894-231712894 on Build GRCh38
rs75345231 Chr.2: 231712711-231712711 on Build GRCh38
rs868769422 Chr.2: 231712693-231712693 on Build GRCh38
rs769950396 Chr.2: 231712826-231712826 on Build GRCh38
rs368863947 Chr.2: 231712876-231712877 on Build GRCh38
rs372241948 Chr.2: 231712703-231712703 on Build GRCh38
rs983830836 Chr.2: 231712835-231712835 on Build GRCh38
rs999579085 Chr.2: 231712749-231712749 on Build GRCh38

View Full Product Details