Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160353039-160353280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
UniGene
Hs.517249
Gene Chromosome Location
Chr.1: 160343056-160358952 on Build GRCh38
Transcripts
NM_001290184.1 NM_001290186.1 NM_015331.2 XM_005245053.4 XM_011509363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770795 c.1119+8G>C p.? Chr.1: 160353245-160353245 on Build GRCh38
4819697 c.1144C>T p.P382S Chr.1: 160353202-160353202 on Build GRCh38
386157 c.1155G>A p.Q385Q Chr.1: 160353213-160353213 on Build GRCh38
6283809 c.1135C>T p.H379Y Chr.1: 160353193-160353193 on Build GRCh38
6283810 c.1075C>T p.H359Y Chr.1: 160353193-160353193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920971774 Chr.1: 160353160-160353160 on Build GRCh38
rs199659075 Chr.1: 160353111-160353111 on Build GRCh38
rs764288790 Chr.1: 160353178-160353178 on Build GRCh38
rs544552209 Chr.1: 160353249-160353249 on Build GRCh38
rs746999382 Chr.1: 160353235-160353235 on Build GRCh38
rs760016375 Chr.1: 160353153-160353153 on Build GRCh38
rs10158574 Chr.1: 160353064-160353064 on Build GRCh38
rs376909952 Chr.1: 160353156-160353156 on Build GRCh38
rs200815363 Chr.1: 160353171-160353171 on Build GRCh38
rs746897539 Chr.1: 160353136-160353136 on Build GRCh38
rs534122547 Chr.1: 160353076-160353076 on Build GRCh38
rs771760093 Chr.1: 160353130-160353130 on Build GRCh38
rs568248629 Chr.1: 160353075-160353075 on Build GRCh38
rs768193810 Chr.1: 160353142-160353142 on Build GRCh38
rs759557582 Chr.1: 160353151-160353152 on Build GRCh38
rs988122193 Chr.1: 160353091-160353091 on Build GRCh38
rs199830369 Chr.1: 160353224-160353224 on Build GRCh38
rs553747081 Chr.1: 160353196-160353196 on Build GRCh38
rs375632985 Chr.1: 160353150-160353150 on Build GRCh38
rs757553805 Chr.1: 160353226-160353226 on Build GRCh38
rs374951668 Chr.1: 160353230-160353230 on Build GRCh38
rs1033640155 Chr.1: 160353077-160353077 on Build GRCh38
rs779680771 Chr.1: 160353233-160353233 on Build GRCh38
rs151131279 Chr.1: 160353229-160353229 on Build GRCh38
rs769379850 Chr.1: 160353152-160353152 on Build GRCh38
rs767666512 Chr.1: 160353152-160353154 on Build GRCh38
rs866536656 Chr.1: 160353155-160353155 on Build GRCh38
rs926453200 Chr.1: 160353220-160353220 on Build GRCh38
rs761044931 Chr.1: 160353157-160353157 on Build GRCh38
rs780096348 Chr.1: 160353135-160353135 on Build GRCh38
rs1000460714 Chr.1: 160353066-160353066 on Build GRCh38
rs201642962 Chr.1: 160353177-160353177 on Build GRCh38

View Full Product Details