Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49636478-49636741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAS
Entrez Gene ID
6237
Gene Name
related RAS viral (r-ras) oncogene homolog
Gene Aliases
-
UniGene
Hs.515536
Gene Chromosome Location
Chr.19: 49635292-49640143 on Build GRCh38
Transcripts
NM_006270.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2752951 c.409G>A p.V137I Chr.19: 49636663-49636663 on Build GRCh38
999356 c.400G>A p.D134N Chr.19: 49636672-49636672 on Build GRCh38
3835388 c.396C>T p.R132R Chr.19: 49636676-49636676 on Build GRCh38
999355 c.418G>T p.V140F Chr.19: 49636654-49636654 on Build GRCh38
2752950 c.423G>T p.G141G Chr.19: 49636649-49636649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775018539 Chr.19: 49636649-49636649 on Build GRCh38
rs117957448 Chr.19: 49636599-49636599 on Build GRCh38
rs377252429 Chr.19: 49636578-49636578 on Build GRCh38
rs374077706 Chr.19: 49636721-49636721 on Build GRCh38
rs964898803 Chr.19: 49636626-49636626 on Build GRCh38
rs114187560 Chr.19: 49636664-49636664 on Build GRCh38
rs767209383 Chr.19: 49636624-49636624 on Build GRCh38
rs749228857 Chr.19: 49636579-49636579 on Build GRCh38
rs757157570 Chr.19: 49636580-49636580 on Build GRCh38
rs745598358 Chr.19: 49636585-49636585 on Build GRCh38
rs974724531 Chr.19: 49636687-49636687 on Build GRCh38
rs776651282 Chr.19: 49636592-49636592 on Build GRCh38
rs896071517 Chr.19: 49636645-49636645 on Build GRCh38
rs138124318 Chr.19: 49636710-49636710 on Build GRCh38
rs557007484 Chr.19: 49636709-49636709 on Build GRCh38
rs373485975 Chr.19: 49636651-49636651 on Build GRCh38
rs372935035 Chr.19: 49636511-49636511 on Build GRCh38
rs749320172 Chr.19: 49636689-49636689 on Build GRCh38
rs368194123 Chr.19: 49636582-49636582 on Build GRCh38
rs1009331090 Chr.19: 49636584-49636584 on Build GRCh38
rs368978550 Chr.19: 49636700-49636700 on Build GRCh38
rs374650566 Chr.19: 49636672-49636672 on Build GRCh38
rs142277653 Chr.19: 49636677-49636677 on Build GRCh38
rs759782098 Chr.19: 49636617-49636617 on Build GRCh38
rs533583599 Chr.19: 49636720-49636720 on Build GRCh38
rs766707816 Chr.19: 49636614-49636614 on Build GRCh38
rs372572482 Chr.19: 49636576-49636576 on Build GRCh38
rs773775763 Chr.19: 49636608-49636608 on Build GRCh38
rs369240501 Chr.19: 49636650-49636650 on Build GRCh38
rs376560391 Chr.19: 49636613-49636613 on Build GRCh38
rs753845212 Chr.19: 49636597-49636597 on Build GRCh38
rs369508242 Chr.19: 49636701-49636701 on Build GRCh38
rs1056225304 Chr.19: 49636517-49636517 on Build GRCh38
rs757080959 Chr.19: 49636663-49636663 on Build GRCh38
rs767030200 Chr.19: 49636573-49636573 on Build GRCh38
rs773258112 Chr.19: 49636678-49636678 on Build GRCh38
rs61760904 Chr.19: 49636675-49636675 on Build GRCh38
rs774660560 Chr.19: 49636697-49636697 on Build GRCh38
rs565545701 Chr.19: 49636516-49636516 on Build GRCh38
rs145907892 Chr.19: 49636693-49636693 on Build GRCh38
rs372053161 Chr.19: 49636591-49636591 on Build GRCh38
rs374239300 Chr.19: 49636521-49636521 on Build GRCh38
rs199656328 Chr.19: 49636705-49636705 on Build GRCh38
rs748366671 Chr.19: 49636676-49636676 on Build GRCh38
rs576507390 Chr.19: 49636652-49636652 on Build GRCh38
rs752382513 Chr.19: 49636631-49636631 on Build GRCh38
rs760231494 Chr.19: 49636523-49636523 on Build GRCh38
rs774803334 Chr.19: 49636615-49636615 on Build GRCh38
rs1017555483 Chr.19: 49636623-49636623 on Build GRCh38
rs758071792 Chr.19: 49636673-49636673 on Build GRCh38
rs769850874 Chr.19: 49636605-49636605 on Build GRCh38

View Full Product Details