Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 106359497-106359770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXR1
Entrez Gene ID
55074
Gene Name
oxidation resistance 1
Gene Aliases
Nbla00307, TLDC3
UniGene
Hs.127286
Gene Chromosome Location
Chr.8: 106270178-106752694 on Build GRCh38
Transcripts
NM_001198533.1 NM_018002.3 XM_017013596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4471104 c.16C>T p.L6L Chr.8: 106359629-106359629 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2022982 Chr.8: 106359696-106359696 on Build GRCh38
rs887655909 Chr.8: 106359550-106359550 on Build GRCh38
rs183590975 Chr.8: 106359552-106359552 on Build GRCh38
rs367686247 Chr.8: 106359565-106359565 on Build GRCh38
rs980558390 Chr.8: 106359542-106359542 on Build GRCh38
rs761428117 Chr.8: 106359726-106359726 on Build GRCh38
rs1030478790 Chr.8: 106359595-106359595 on Build GRCh38
rs781398630 Chr.8: 106359646-106359646 on Build GRCh38
rs1035998356 Chr.8: 106359574-106359574 on Build GRCh38
rs371568162 Chr.8: 106359543-106359543 on Build GRCh38
rs997212996 Chr.8: 106359588-106359588 on Build GRCh38
rs368917619 Chr.8: 106359609-106359609 on Build GRCh38
rs867488400 Chr.8: 106359739-106359739 on Build GRCh38
rs897500352 Chr.8: 106359578-106359578 on Build GRCh38
rs143910836 Chr.8: 106359702-106359702 on Build GRCh38
rs1005894200 Chr.8: 106359585-106359585 on Build GRCh38
rs914613464 Chr.8: 106359538-106359538 on Build GRCh38
rs1055591867 Chr.8: 106359544-106359544 on Build GRCh38
rs982168650 Chr.8: 106359711-106359711 on Build GRCh38
rs988812839 Chr.8: 106359703-106359703 on Build GRCh38
rs888904972 Chr.8: 106359663-106359663 on Build GRCh38
rs971118240 Chr.8: 106359709-106359709 on Build GRCh38
rs773963389 Chr.8: 106359723-106359723 on Build GRCh38
rs754276919 Chr.8: 106359665-106359665 on Build GRCh38
rs771298080 Chr.8: 106359611-106359611 on Build GRCh38
rs566957296 Chr.8: 106359535-106359535 on Build GRCh38
rs1047427749 Chr.8: 106359556-106359556 on Build GRCh38
rs938099220 Chr.8: 106359738-106359738 on Build GRCh38
rs375881128 Chr.8: 106359599-106359599 on Build GRCh38
rs1007731497 Chr.8: 106359666-106359666 on Build GRCh38
rs117341867 Chr.8: 106359707-106359707 on Build GRCh38
rs947410847 Chr.8: 106359541-106359541 on Build GRCh38

View Full Product Details