Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10643681-10643954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAG1
Entrez Gene ID
182
Gene Name
jagged 1
Gene Aliases
AGS, AHD, AWS, CD339, HJ1, JAGL1
UniGene
Hs.224012
Gene Chromosome Location
Chr.20: 10637684-10674046 on Build GRCh38
Transcripts
NM_000214.2 XM_017027707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6689211 c.2450G>A p.C817Y Chr.20: 10643786-10643786 on Build GRCh38
1024439 c.2419G>A p.E807K Chr.20: 10643817-10643817 on Build GRCh38
4537257 c.2399G>A p.G800E Chr.20: 10643837-10643837 on Build GRCh38
3544074 c.2442G>A p.G814G Chr.20: 10643794-10643794 on Build GRCh38
5880132 c.2413C>T p.R805W Chr.20: 10643823-10643823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771297154 Chr.20: 10643892-10643892 on Build GRCh38
rs779762642 Chr.20: 10643881-10643881 on Build GRCh38
rs747532570 Chr.20: 10643785-10643785 on Build GRCh38
rs566557171 Chr.20: 10643870-10643870 on Build GRCh38
rs763720023 Chr.20: 10643821-10643821 on Build GRCh38
rs957553735 Chr.20: 10643894-10643895 on Build GRCh38
rs201781026 Chr.20: 10643883-10643883 on Build GRCh38
rs751577946 Chr.20: 10643733-10643733 on Build GRCh38
rs946451108 Chr.20: 10643879-10643879 on Build GRCh38
rs778204780 Chr.20: 10643768-10643768 on Build GRCh38
rs865828029 Chr.20: 10643855-10643855 on Build GRCh38
rs146155406 Chr.20: 10643716-10643716 on Build GRCh38
rs987799112 Chr.20: 10643853-10643853 on Build GRCh38
rs756949835 Chr.20: 10643735-10643735 on Build GRCh38
rs752939939 Chr.20: 10643891-10643891 on Build GRCh38
rs150759762 Chr.20: 10643840-10643840 on Build GRCh38
rs542572569 Chr.20: 10643737-10643737 on Build GRCh38
rs769531968 Chr.20: 10643807-10643807 on Build GRCh38
rs777122869 Chr.20: 10643791-10643791 on Build GRCh38
rs533306015 Chr.20: 10643818-10643818 on Build GRCh38
rs761813198 Chr.20: 10643839-10643839 on Build GRCh38
rs56225585 Chr.20: 10643854-10643854 on Build GRCh38
rs748268587 Chr.20: 10643756-10643756 on Build GRCh38
rs564566567 Chr.20: 10643817-10643817 on Build GRCh38
rs876661165 Chr.20: 10643818-10643818 on Build GRCh38
rs745496042 Chr.20: 10643794-10643794 on Build GRCh38
rs372547927 Chr.20: 10643895-10643895 on Build GRCh38
rs77308315 Chr.20: 10643750-10643750 on Build GRCh38
rs772170740 Chr.20: 10643762-10643762 on Build GRCh38
rs199571405 Chr.20: 10643738-10643738 on Build GRCh38
rs759194615 Chr.20: 10643726-10643726 on Build GRCh38
rs372199268 Chr.20: 10643727-10643727 on Build GRCh38
rs776080544 Chr.20: 10643719-10643719 on Build GRCh38
rs2273059 Chr.20: 10643748-10643748 on Build GRCh38
rs374941347 Chr.20: 10643790-10643790 on Build GRCh38
rs774372899 Chr.20: 10643822-10643822 on Build GRCh38

View Full Product Details