Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122510828-122511087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR1J2
Entrez Gene ID
26740
Gene Name
olfactory receptor family 1 subfamily J member 2
Gene Aliases
HG152, HSA5, OR1J3, OR1J5, OR9-19, OST044
UniGene
Hs.460145
Gene Chromosome Location
Chr.9: 122403407-122514097 on Build GRCh38
Transcripts
NM_054107.1 XM_017014623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7381267 c.191G>T p.S64I Chr.9: 122510992-122510992 on Build GRCh38
3323365 c.244C>A p.L82M Chr.9: 122511045-122511045 on Build GRCh38
4709692 c.271T>G p.S91A Chr.9: 122511072-122511072 on Build GRCh38
3779604 c.158C>G p.S53C Chr.9: 122510959-122510959 on Build GRCh38
607456 c.272C>T p.S91L Chr.9: 122511073-122511073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367711559 Chr.9: 122511043-122511043 on Build GRCh38
rs765101019 Chr.9: 122510972-122510972 on Build GRCh38
rs754640814 Chr.9: 122511021-122511021 on Build GRCh38
rs559485689 Chr.9: 122510902-122510902 on Build GRCh38
rs771628528 Chr.9: 122510936-122510936 on Build GRCh38
rs150299329 Chr.9: 122510974-122510974 on Build GRCh38
rs201738524 Chr.9: 122510900-122510900 on Build GRCh38
rs772347154 Chr.9: 122510954-122510954 on Build GRCh38
rs760385807 Chr.9: 122511045-122511045 on Build GRCh38
rs746189490 Chr.9: 122510948-122510948 on Build GRCh38
rs201234508 Chr.9: 122511028-122511028 on Build GRCh38
rs749849611 Chr.9: 122510854-122510854 on Build GRCh38
rs780333028 Chr.9: 122511050-122511050 on Build GRCh38
rs137857788 Chr.9: 122510992-122510992 on Build GRCh38
rs201746798 Chr.9: 122510914-122510914 on Build GRCh38
rs978663736 Chr.9: 122510866-122510866 on Build GRCh38
rs142519029 Chr.9: 122510876-122510876 on Build GRCh38
rs138188243 Chr.9: 122511059-122511059 on Build GRCh38
rs150403833 Chr.9: 122510924-122510924 on Build GRCh38
rs576241056 Chr.9: 122510964-122510964 on Build GRCh38
rs747242295 Chr.9: 122511052-122511052 on Build GRCh38
rs768934378 Chr.9: 122511054-122511054 on Build GRCh38
rs560541233 Chr.9: 122511056-122511056 on Build GRCh38
rs774947810 Chr.9: 122511031-122511031 on Build GRCh38
rs779515823 Chr.9: 122511035-122511035 on Build GRCh38
rs751200672 Chr.9: 122510906-122510906 on Build GRCh38
rs750151678 Chr.9: 122510895-122510895 on Build GRCh38
rs200087305 Chr.9: 122510855-122510855 on Build GRCh38
rs769891533 Chr.9: 122511058-122511058 on Build GRCh38
rs767716131 Chr.9: 122510860-122510860 on Build GRCh38
rs560859704 Chr.9: 122510935-122510935 on Build GRCh38
rs142471257 Chr.9: 122510896-122510896 on Build GRCh38
rs751200072 Chr.9: 122511003-122511003 on Build GRCh38
rs753745594 Chr.9: 122510887-122510887 on Build GRCh38
rs779571149 Chr.9: 122510939-122510939 on Build GRCh38
rs200778732 Chr.9: 122510859-122510859 on Build GRCh38
rs546772045 Chr.9: 122510857-122510857 on Build GRCh38
rs765004628 Chr.9: 122510894-122510894 on Build GRCh38
rs761886029 Chr.9: 122510971-122510971 on Build GRCh38
rs992535520 Chr.9: 122510849-122510849 on Build GRCh38
rs867442844 Chr.9: 122511025-122511025 on Build GRCh38
rs183379346 Chr.9: 122511011-122511011 on Build GRCh38
rs151331246 Chr.9: 122510915-122510915 on Build GRCh38
rs766124858 Chr.9: 122510996-122510996 on Build GRCh38
rs370861596 Chr.9: 122510877-122510877 on Build GRCh38
rs201194266 Chr.9: 122510961-122510961 on Build GRCh38
rs757234772 Chr.9: 122510889-122510889 on Build GRCh38
rs755703163 Chr.9: 122511033-122511033 on Build GRCh38
rs762767361 Chr.9: 122510994-122510994 on Build GRCh38
rs141998750 Chr.9: 122510874-122510874 on Build GRCh38
rs920954562 Chr.9: 122510988-122510988 on Build GRCh38
rs780896253 Chr.9: 122510932-122510932 on Build GRCh38
rs761831251 Chr.9: 122511057-122511057 on Build GRCh38
rs780841335 Chr.9: 122511023-122511023 on Build GRCh38

View Full Product Details