Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19344736-19345009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAU2
Entrez Gene ID
23383
Gene Name
MAU2 sister chromatid cohesion factor
Gene Aliases
KIAA0892, MAU2L, SCC4, mau-2
UniGene
Hs.654939
Gene Chromosome Location
Chr.19: 19320686-19358754 on Build GRCh38
Transcripts
NM_015329.3 XM_005259837.3 XM_005259838.3 XM_006722711.3 XM_011527844.2 XM_017026539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769402 c.1087G>A p.V363I Chr.19: 19344858-19344858 on Build GRCh38
5855142 c.1133C>T p.A378V Chr.19: 19344904-19344904 on Build GRCh38
3101256 c.1109delC p.R372fs*115 Chr.19: 19344880-19344880 on Build GRCh38
5389167 c.1149A>T p.T383T Chr.19: 19344920-19344920 on Build GRCh38
4075927 c.1115G>T p.R372L Chr.19: 19344886-19344886 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374562642 Chr.19: 19344842-19344842 on Build GRCh38
rs988960135 Chr.19: 19344797-19344797 on Build GRCh38
rs140913237 Chr.19: 19344844-19344844 on Build GRCh38
rs771048411 Chr.19: 19344895-19344895 on Build GRCh38
rs762910396 Chr.19: 19344918-19344918 on Build GRCh38
rs944461782 Chr.19: 19344759-19344759 on Build GRCh38
rs370992314 Chr.19: 19344866-19344866 on Build GRCh38
rs758772268 Chr.19: 19344878-19344878 on Build GRCh38
rs776644565 Chr.19: 19344897-19344897 on Build GRCh38
rs968584180 Chr.19: 19344810-19344810 on Build GRCh38
rs779363857 Chr.19: 19344858-19344858 on Build GRCh38
rs751386140 Chr.19: 19344929-19344929 on Build GRCh38
rs769822298 Chr.19: 19344946-19344946 on Build GRCh38
rs200693842 Chr.19: 19344951-19344951 on Build GRCh38
rs763874426 Chr.19: 19344921-19344921 on Build GRCh38
rs1002171183 Chr.19: 19344887-19344887 on Build GRCh38
rs552627506 Chr.19: 19344880-19344880 on Build GRCh38
rs60890167 Chr.19: 19344937-19344937 on Build GRCh38
rs761574732 Chr.19: 19344933-19344933 on Build GRCh38
rs776697682 Chr.19: 19344967-19344967 on Build GRCh38
rs751724598 Chr.19: 19344973-19344976 on Build GRCh38
rs779730747 Chr.19: 19344945-19344945 on Build GRCh38
rs374459100 Chr.19: 19344952-19344952 on Build GRCh38
rs8102280 Chr.19: 19344941-19344941 on Build GRCh38
rs755756207 Chr.19: 19344807-19344807 on Build GRCh38
rs986801450 Chr.19: 19344885-19344885 on Build GRCh38
rs903827673 Chr.19: 19344845-19344845 on Build GRCh38
rs747254397 Chr.19: 19344953-19344953 on Build GRCh38
rs745893860 Chr.19: 19344970-19344970 on Build GRCh38
rs771103746 Chr.19: 19344964-19344964 on Build GRCh38
rs367748761 Chr.19: 19344972-19344972 on Build GRCh38
rs750081178 Chr.19: 19344798-19344798 on Build GRCh38
rs1040228631 Chr.19: 19344761-19344761 on Build GRCh38
rs371904513 Chr.19: 19344882-19344882 on Build GRCh38
rs1033668015 Chr.19: 19344920-19344920 on Build GRCh38
rs755544796 Chr.19: 19344948-19344948 on Build GRCh38
rs577947201 Chr.19: 19344985-19344985 on Build GRCh38
rs779675886 Chr.19: 19344843-19344843 on Build GRCh38

View Full Product Details