Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54178714-54178987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBOAT7
Entrez Gene ID
79143
Gene Name
membrane bound O-acyltransferase domain containing 7
Gene Aliases
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7
UniGene
Hs.467279
Gene Chromosome Location
Chr.19: 54173412-54190050 on Build GRCh38
Transcripts
NM_001146056.2 NM_001146082.2 NM_001146083.2 NM_024298.4 XM_011527299.2 XM_011527300.2 XM_017027296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1001026 c.990G>T p.Q330H Chr.19: 54178806-54178806 on Build GRCh38
312777 c.1015delC p.R339fs*13 Chr.19: 54178781-54178781 on Build GRCh38
6356460 c.931G>A p.V311M Chr.19: 54178865-54178865 on Build GRCh38
1001027 c.976T>C p.W326R Chr.19: 54178820-54178820 on Build GRCh38
5744010 c.968C>T p.T323M Chr.19: 54178828-54178828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766171529 Chr.19: 54178920-54178920 on Build GRCh38
rs757489929 Chr.19: 54178939-54178939 on Build GRCh38
rs780802938 Chr.19: 54178744-54178744 on Build GRCh38
rs761264534 Chr.19: 54178780-54178780 on Build GRCh38
rs369722870 Chr.19: 54178753-54178753 on Build GRCh38
rs766981578 Chr.19: 54178910-54178910 on Build GRCh38
rs771446633 Chr.19: 54178869-54178869 on Build GRCh38
rs771590260 Chr.19: 54178964-54178964 on Build GRCh38
rs879020029 Chr.19: 54178933-54178933 on Build GRCh38
rs962176750 Chr.19: 54178813-54178813 on Build GRCh38
rs762596416 Chr.19: 54178784-54178784 on Build GRCh38
rs145343446 Chr.19: 54178898-54178898 on Build GRCh38
rs574129950 Chr.19: 54178961-54178961 on Build GRCh38
rs761899500 Chr.19: 54178765-54178765 on Build GRCh38
rs753794442 Chr.19: 54178856-54178856 on Build GRCh38
rs769652995 Chr.19: 54178749-54178749 on Build GRCh38
rs201042146 Chr.19: 54178932-54178932 on Build GRCh38
rs767055612 Chr.19: 54178781-54178781 on Build GRCh38
rs1049872070 Chr.19: 54178844-54178844 on Build GRCh38
rs377603361 Chr.19: 54178855-54178855 on Build GRCh38
rs761381990 Chr.19: 54178849-54178849 on Build GRCh38
rs373319764 Chr.19: 54178853-54178853 on Build GRCh38
rs749428006 Chr.19: 54178752-54178752 on Build GRCh38
rs376378590 Chr.19: 54178809-54178809 on Build GRCh38
rs745820347 Chr.19: 54178745-54178745 on Build GRCh38
rs756804673 Chr.19: 54178791-54178791 on Build GRCh38
rs760053104 Chr.19: 54178745-54178748 on Build GRCh38
rs770174442 Chr.19: 54178957-54178957 on Build GRCh38
rs760271247 Chr.19: 54178919-54178919 on Build GRCh38
rs758487661 Chr.19: 54178862-54178862 on Build GRCh38
rs750286516 Chr.19: 54178803-54178803 on Build GRCh38
rs776052515 Chr.19: 54178892-54178892 on Build GRCh38
rs369198461 Chr.19: 54178865-54178865 on Build GRCh38
rs773559537 Chr.19: 54178824-54178824 on Build GRCh38
rs765564429 Chr.19: 54178776-54178776 on Build GRCh38
rs146777664 Chr.19: 54178911-54178911 on Build GRCh38
rs755890292 Chr.19: 54178804-54178804 on Build GRCh38
rs781562749 Chr.19: 54178948-54178948 on Build GRCh38
rs377399161 Chr.19: 54178832-54178832 on Build GRCh38
rs139499369 Chr.19: 54178828-54178828 on Build GRCh38
rs778806571 Chr.19: 54178820-54178820 on Build GRCh38
rs757972919 Chr.19: 54178736-54178736 on Build GRCh38
rs746276330 Chr.19: 54178954-54178954 on Build GRCh38
rs747431480 Chr.19: 54178827-54178827 on Build GRCh38
rs543413097 Chr.19: 54178930-54178930 on Build GRCh38
rs369686836 Chr.19: 54178842-54178842 on Build GRCh38
rs757108954 Chr.19: 54178740-54178740 on Build GRCh38
rs1034934225 Chr.19: 54178748-54178748 on Build GRCh38
rs773651355 Chr.19: 54178779-54178779 on Build GRCh38
rs149699444 Chr.19: 54178866-54178866 on Build GRCh38
rs143957744 Chr.19: 54178810-54178810 on Build GRCh38
rs780899590 Chr.19: 54178951-54178951 on Build GRCh38
rs748238566 Chr.19: 54178861-54178861 on Build GRCh38
rs202020450 Chr.19: 54178790-54178790 on Build GRCh38
rs888670929 Chr.19: 54178966-54178966 on Build GRCh38
rs139422301 Chr.19: 54178797-54178797 on Build GRCh38
rs753507465 Chr.19: 54178922-54178922 on Build GRCh38
rs112211751 Chr.19: 54178763-54178763 on Build GRCh38
rs755943204 Chr.19: 54178854-54178854 on Build GRCh38
rs753089005 Chr.19: 54178925-54178974 on Build GRCh38
rs751847726 Chr.19: 54178938-54178938 on Build GRCh38
rs116423748 Chr.19: 54178821-54178821 on Build GRCh38
rs773670226 Chr.19: 54178847-54178847 on Build GRCh38
rs754793229 Chr.19: 54178929-54178929 on Build GRCh38
rs200105516 Chr.19: 54178766-54178766 on Build GRCh38
rs763676201 Chr.19: 54178843-54178843 on Build GRCh38
rs534942399 Chr.19: 54178860-54178860 on Build GRCh38
rs952247185 Chr.19: 54178802-54178802 on Build GRCh38
rs557746011 Chr.19: 54178889-54178889 on Build GRCh38
rs777081257 Chr.19: 54178887-54178887 on Build GRCh38
rs373026642 Chr.19: 54178931-54178931 on Build GRCh38
rs754379538 Chr.19: 54178783-54178783 on Build GRCh38
rs756923309 Chr.19: 54178943-54178944 on Build GRCh38
rs774672607 Chr.19: 54178757-54178757 on Build GRCh38
rs777534486 Chr.19: 54178738-54178738 on Build GRCh38
rs772115812 Chr.19: 54178823-54178823 on Build GRCh38
rs372356292 Chr.19: 54178786-54178786 on Build GRCh38
rs749792902 Chr.19: 54178945-54178945 on Build GRCh38

View Full Product Details