Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140803530-140803787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5931704 c.2380G>A p.D794N Chr.5: 140803577-140803577 on Build GRCh38
4843003 c.2438C>G p.S813* Chr.5: 140803635-140803635 on Build GRCh38
6469990 c.2343T>A p.I781I Chr.5: 140803540-140803540 on Build GRCh38
1433774 c.2420C>A p.P807Q Chr.5: 140803617-140803617 on Build GRCh38
6500465 c.2403delT p.Y803fs*>22 Chr.5: 140803600-140803600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782622093 Chr.5: 140803557-140803557 on Build GRCh38
rs781820342 Chr.5: 140803678-140803678 on Build GRCh38
rs543206903 Chr.5: 140803726-140803726 on Build GRCh38
rs782142850 Chr.5: 140803638-140803638 on Build GRCh38
rs577997860 Chr.5: 140803612-140803612 on Build GRCh38
rs376686641 Chr.5: 140803590-140803590 on Build GRCh38
rs782270719 Chr.5: 140803593-140803593 on Build GRCh38
rs782685933 Chr.5: 140803627-140803627 on Build GRCh38
rs782640971 Chr.5: 140803686-140803686 on Build GRCh38
rs781914868 Chr.5: 140803698-140803698 on Build GRCh38
rs17844265 Chr.5: 140803652-140803652 on Build GRCh38
rs782061277 Chr.5: 140803625-140803625 on Build GRCh38
rs782003292 Chr.5: 140803708-140803708 on Build GRCh38
rs201656415 Chr.5: 140803607-140803607 on Build GRCh38
rs1003396292 Chr.5: 140803622-140803622 on Build GRCh38
rs974545602 Chr.5: 140803703-140803703 on Build GRCh38
rs2240693 Chr.5: 140803740-140803740 on Build GRCh38
rs782316183 Chr.5: 140803693-140803693 on Build GRCh38
rs782421319 Chr.5: 140803589-140803589 on Build GRCh38
rs781853689 Chr.5: 140803644-140803644 on Build GRCh38
rs782237100 Chr.5: 140803690-140803690 on Build GRCh38
rs7707310 Chr.5: 140803680-140803680 on Build GRCh38
rs898664223 Chr.5: 140803569-140803569 on Build GRCh38
rs941737742 Chr.5: 140803755-140803755 on Build GRCh38
rs782283652 Chr.5: 140803702-140803702 on Build GRCh38
rs539044663 Chr.5: 140803649-140803649 on Build GRCh38
rs782182345 Chr.5: 140803587-140803587 on Build GRCh38
rs782597617 Chr.5: 140803578-140803578 on Build GRCh38
rs782118084 Chr.5: 140803714-140803714 on Build GRCh38
rs190117993 Chr.5: 140803707-140803707 on Build GRCh38
rs782224885 Chr.5: 140803562-140803562 on Build GRCh38
rs782466651 Chr.5: 140803572-140803572 on Build GRCh38
rs781810632 Chr.5: 140803653-140803653 on Build GRCh38
rs572043416 Chr.5: 140803675-140803675 on Build GRCh38
rs782450131 Chr.5: 140803671-140803671 on Build GRCh38
rs560789364 Chr.5: 140803683-140803683 on Build GRCh38
rs182945871 Chr.5: 140803739-140803739 on Build GRCh38

View Full Product Details