Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171512980-171513205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
UniGene
Hs.494614
Gene Chromosome Location
Chr.1: 171485496-171593511 on Build GRCh38
Transcripts
NM_015172.3 XM_005245015.2 XM_005245016.4 XM_005245017.2 XM_005245018.2 XM_005245019.2 XM_005245020.2 XM_005245023.2 XM_005245024.2 XM_005245025.2 XM_005245026.2 XM_005245027.2 XM_005245028.2 XM_005245029.2 XM_006711238.3 XM_017000760.1 XM_017000761.1 XM_017000762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899523 c.122G>A p.R41Q Chr.1: 171513004-171513004 on Build GRCh38
899525 c.139C>A p.L47I Chr.1: 171513021-171513021 on Build GRCh38
6757039 c.151G>A p.G51S Chr.1: 171513033-171513033 on Build GRCh38
899527 c.148G>A p.V50I Chr.1: 171513030-171513030 on Build GRCh38
899529 c.163C>T p.R55C Chr.1: 171513045-171513045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760011288 Chr.1: 171513113-171513114 on Build GRCh38
rs376186276 Chr.1: 171513126-171513126 on Build GRCh38
rs371016782 Chr.1: 171513097-171513097 on Build GRCh38
rs763548975 Chr.1: 171513127-171513127 on Build GRCh38
rs774065990 Chr.1: 171513043-171513043 on Build GRCh38
rs766175434 Chr.1: 171513083-171513083 on Build GRCh38
rs377615253 Chr.1: 171513143-171513143 on Build GRCh38
rs778001715 Chr.1: 171513155-171513155 on Build GRCh38
rs576045141 Chr.1: 171513068-171513068 on Build GRCh38
rs754976574 Chr.1: 171513100-171513100 on Build GRCh38
rs901423141 Chr.1: 171513141-171513141 on Build GRCh38
rs1041318971 Chr.1: 171513113-171513113 on Build GRCh38
rs760121094 Chr.1: 171513036-171513036 on Build GRCh38
rs35546712 Chr.1: 171513023-171513024 on Build GRCh38
rs138939414 Chr.1: 171513032-171513032 on Build GRCh38
rs763689482 Chr.1: 171513042-171513042 on Build GRCh38
rs777218200 Chr.1: 171513034-171513034 on Build GRCh38
rs533213461 Chr.1: 171513071-171513071 on Build GRCh38
rs761353772 Chr.1: 171513053-171513053 on Build GRCh38
rs942687278 Chr.1: 171513109-171513109 on Build GRCh38
rs748298563 Chr.1: 171513136-171513136 on Build GRCh38
rs747206117 Chr.1: 171513160-171513160 on Build GRCh38
rs767242003 Chr.1: 171513056-171513056 on Build GRCh38
rs750077680 Chr.1: 171513065-171513065 on Build GRCh38

View Full Product Details