Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112918384-112918637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL37
Entrez Gene ID
27178
Gene Name
interleukin 37
Gene Aliases
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1
UniGene
Hs.166371
Gene Chromosome Location
Chr.2: 112908889-112918882 on Build GRCh38
Transcripts
NM_014439.3 NM_173202.1 NM_173203.1 NM_173204.1 NM_173205.1 XM_011510963.2 XM_011510964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
714713 c.365A>G p.E122G Chr.2: 112918595-112918595 on Build GRCh38
4952173 c.355G>A p.A119T Chr.2: 112918585-112918585 on Build GRCh38
714715 c.342G>A p.L114L Chr.2: 112918572-112918572 on Build GRCh38
3040033 c.447A>G p.S149S Chr.2: 112918599-112918599 on Build GRCh38
3040034 c.369A>G p.S123S Chr.2: 112918599-112918599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555978965 Chr.2: 112918585-112918585 on Build GRCh38
rs779931904 Chr.2: 112918486-112918486 on Build GRCh38
rs779607633 Chr.2: 112918510-112918510 on Build GRCh38
rs536123660 Chr.2: 112918570-112918570 on Build GRCh38
rs373240605 Chr.2: 112918553-112918553 on Build GRCh38
rs144279173 Chr.2: 112918610-112918610 on Build GRCh38
rs759647208 Chr.2: 112918579-112918579 on Build GRCh38
rs978225478 Chr.2: 112918463-112918463 on Build GRCh38
rs911234266 Chr.2: 112918527-112918527 on Build GRCh38
rs774335153 Chr.2: 112918571-112918571 on Build GRCh38
rs566074324 Chr.2: 112918445-112918445 on Build GRCh38
rs924533688 Chr.2: 112918421-112918421 on Build GRCh38
rs776222274 Chr.2: 112918546-112918546 on Build GRCh38
rs547310623 Chr.2: 112918480-112918480 on Build GRCh38
rs756247588 Chr.2: 112918589-112918589 on Build GRCh38
rs934054389 Chr.2: 112918484-112918484 on Build GRCh38
rs1047867137 Chr.2: 112918599-112918599 on Build GRCh38
rs746524603 Chr.2: 112918536-112918536 on Build GRCh38
rs532660211 Chr.2: 112918603-112918603 on Build GRCh38
rs986842042 Chr.2: 112918525-112918525 on Build GRCh38
rs28947200 Chr.2: 112918606-112918606 on Build GRCh38
rs569745618 Chr.2: 112918604-112918604 on Build GRCh38
rs866423799 Chr.2: 112918563-112918563 on Build GRCh38
rs915907545 Chr.2: 112918450-112918450 on Build GRCh38
rs145006297 Chr.2: 112918425-112918425 on Build GRCh38
rs534674855 Chr.2: 112918470-112918470 on Build GRCh38
rs772939360 Chr.2: 112918555-112918555 on Build GRCh38
rs377488590 Chr.2: 112918607-112918607 on Build GRCh38
rs552343137 Chr.2: 112918444-112918444 on Build GRCh38
rs990119174 Chr.2: 112918435-112918435 on Build GRCh38
rs752825177 Chr.2: 112918586-112918586 on Build GRCh38
rs560375860 Chr.2: 112918548-112918548 on Build GRCh38
rs146771722 Chr.2: 112918538-112918538 on Build GRCh38
rs747692110 Chr.2: 112918549-112918549 on Build GRCh38

View Full Product Details