Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141124043-141124316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB4
Entrez Gene ID
56131
Gene Name
protocadherin beta 4
Gene Aliases
PCDH-BETA4
UniGene
Hs.591250
Gene Chromosome Location
Chr.5: 141121750-141125623 on Build GRCh38
Transcripts
NM_018938.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5867567 c.2267G>A p.G756E Chr.5: 141124265-141124265 on Build GRCh38
6469999 c.2277G>T p.G759G Chr.5: 141124275-141124275 on Build GRCh38
1261389 c.2159C>T p.S720L Chr.5: 141124157-141124157 on Build GRCh38
5785727 c.2058C>A p.T686T Chr.5: 141124056-141124056 on Build GRCh38
1261387 c.2285A>T p.E762V Chr.5: 141124283-141124283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782079186 Chr.5: 141124067-141124067 on Build GRCh38
rs782698921 Chr.5: 141124064-141124064 on Build GRCh38
rs782010525 Chr.5: 141124273-141124273 on Build GRCh38
rs4143454 Chr.5: 141124230-141124230 on Build GRCh38
rs782205928 Chr.5: 141124245-141124245 on Build GRCh38
rs782397979 Chr.5: 141124263-141124263 on Build GRCh38
rs782549930 Chr.5: 141124244-141124244 on Build GRCh38
rs782379004 Chr.5: 141124091-141124091 on Build GRCh38
rs201756242 Chr.5: 141124242-141124242 on Build GRCh38
rs782059174 Chr.5: 141124143-141124143 on Build GRCh38
rs149213680 Chr.5: 141124107-141124107 on Build GRCh38
rs782374322 Chr.5: 141124184-141124184 on Build GRCh38
rs781918620 Chr.5: 141124120-141124120 on Build GRCh38
rs782185764 Chr.5: 141124124-141124124 on Build GRCh38
rs782498865 Chr.5: 141124158-141124158 on Build GRCh38
rs573039653 Chr.5: 141124162-141124162 on Build GRCh38
rs782183136 Chr.5: 141124172-141124172 on Build GRCh38
rs782447575 Chr.5: 141124173-141124173 on Build GRCh38
rs781880373 Chr.5: 141124075-141124075 on Build GRCh38
rs147891724 Chr.5: 141124066-141124066 on Build GRCh38
rs781966778 Chr.5: 141124126-141124126 on Build GRCh38
rs782603101 Chr.5: 141124180-141124180 on Build GRCh38
rs782085452 Chr.5: 141124199-141124199 on Build GRCh38
rs782332407 Chr.5: 141124125-141124125 on Build GRCh38
rs370078570 Chr.5: 141124183-141124183 on Build GRCh38
rs782611297 Chr.5: 141124088-141124088 on Build GRCh38
rs782131892 Chr.5: 141124209-141124209 on Build GRCh38
rs556724572 Chr.5: 141124074-141124074 on Build GRCh38
rs782620651 Chr.5: 141124249-141124249 on Build GRCh38
rs781822336 Chr.5: 141124151-141124151 on Build GRCh38
rs143375135 Chr.5: 141124137-141124137 on Build GRCh38
rs782084409 Chr.5: 141124280-141124280 on Build GRCh38
rs782253966 Chr.5: 141124260-141124260 on Build GRCh38
rs781867029 Chr.5: 141124161-141124161 on Build GRCh38
rs4143455 Chr.5: 141124159-141124159 on Build GRCh38
rs782185062 Chr.5: 141124262-141124262 on Build GRCh38
rs782155638 Chr.5: 141124274-141124274 on Build GRCh38
rs782535078 Chr.5: 141124167-141124167 on Build GRCh38
rs147986368 Chr.5: 141124212-141124212 on Build GRCh38
rs145434045 Chr.5: 141124104-141124104 on Build GRCh38
rs141764271 Chr.5: 141124079-141124079 on Build GRCh38
rs199748518 Chr.5: 141124089-141124089 on Build GRCh38
rs782744230 Chr.5: 141124282-141124282 on Build GRCh38
rs199774224 Chr.5: 141124185-141124185 on Build GRCh38
rs147086302 Chr.5: 141124083-141124083 on Build GRCh38
rs782300686 Chr.5: 141124275-141124275 on Build GRCh38
rs781941387 Chr.5: 141124198-141124198 on Build GRCh38
rs530871407 Chr.5: 141124229-141124229 on Build GRCh38
rs782647381 Chr.5: 141124092-141124092 on Build GRCh38
rs782042589 Chr.5: 141124142-141124142 on Build GRCh38
rs551909584 Chr.5: 141124248-141124248 on Build GRCh38
rs782728424 Chr.5: 141124149-141124149 on Build GRCh38
rs782316970 Chr.5: 141124193-141124193 on Build GRCh38
rs17844419 Chr.5: 141124080-141124080 on Build GRCh38
rs782777687 Chr.5: 141124195-141124195 on Build GRCh38
rs782139580 Chr.5: 141124130-141124130 on Build GRCh38
rs781929599 Chr.5: 141124276-141124276 on Build GRCh38
rs561916518 Chr.5: 141124215-141124215 on Build GRCh38
rs782739668 Chr.5: 141124200-141124200 on Build GRCh38
rs782680472 Chr.5: 141124169-141124169 on Build GRCh38
rs781988555 Chr.5: 141124201-141124201 on Build GRCh38

View Full Product Details