Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 115634834-115635085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLS3
Entrez Gene ID
5358
Gene Name
plastin 3
Gene Aliases
BMND18, T-plastin
UniGene
Hs.496622
Gene Chromosome Location
Chr.X: 115560850-115650861 on Build GRCh38
Transcripts
NM_001136025.4 NM_001172335.2 NM_001282337.1 NM_001282338.1 NM_005032.6 XM_011537534.1

SNP Detail

SNP ID Chromosome Location
rs782192988 Chr.X: 115634934-115634934 on Build GRCh38
rs782008597 Chr.X: 115634988-115634988 on Build GRCh38
rs367830488 Chr.X: 115634938-115634938 on Build GRCh38
rs782582437 Chr.X: 115634904-115634904 on Build GRCh38
rs782227015 Chr.X: 115634900-115634900 on Build GRCh38
rs1048749368 Chr.X: 115634991-115634991 on Build GRCh38
rs371332791 Chr.X: 115634986-115634986 on Build GRCh38
rs781934640 Chr.X: 115635031-115635031 on Build GRCh38
rs781887875 Chr.X: 115635018-115635018 on Build GRCh38
rs782626858 Chr.X: 115634973-115634973 on Build GRCh38
rs781869012 Chr.X: 115634859-115634859 on Build GRCh38
rs782212189 Chr.X: 115635049-115635049 on Build GRCh38
rs375187925 Chr.X: 115635020-115635020 on Build GRCh38
rs782395250 Chr.X: 115634987-115634987 on Build GRCh38
rs894856074 Chr.X: 115634967-115634967 on Build GRCh38
rs112589137 Chr.X: 115634949-115634949 on Build GRCh38

View Full Product Details